Skip to Content
Merck
All Photos(1)

Documents

449644

Sigma-Aldrich

Barium chloride

AnhydroBeads, −10 mesh, 99.999% trace metals basis

Synonym(s):

Barium (2+) dichloride, Barium dichloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
BaCl2
CAS Number:
Molecular Weight:
208.23
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

product line

AnhydroBeads

Assay

99.999% trace metals basis

impurities

≤15.0 ppm Trace Metal Analysis

particle size

−10 mesh

mp

963 °C (lit.)

density

3.856 g/mL at 25 °C (lit.)

SMILES string

Cl[Ba]Cl

InChI

1S/Ba.2ClH/h;2*1H/q+2;;/p-2

InChI key

WDIHJSXYQDMJHN-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Barium chloride (BaCl2) is a water soluble ionic compound that is hygroscopic in nature. It can be prepared from barium hydroxide or barium carbonate and can also be naturally found as witherite.

Application

BaCl2 has a variety of industrial and laboratory applications which include its use as a test for sulfate ion, synthesis of pigments, and manufacturing of pharmaceuticals.

Legal Information

AnhydroBeads is a trademark of Sigma-Aldrich Co. LLC

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Acute Tox. 4 Inhalation - Eye Irrit. 2

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Group 17 (H, F, Cl, Br, I) Alkaline Earth Compounds
Encyclopedia of the Alkaline Earth Compounds, 25-104 (2013)
Role of M 3 receptor in aconitine/barium-chloride-induced preconditioning against arrhythmias in rats
Liu Y, et al.
Naunyn-Schmiedeberg'S Archives of Pharmacology, 379(5), 511-515 (2009)
Alginate-based microencapsulation of retinal pigment epithelial cell line for cell therapy
Wikstrom J, et al.
Biomaterials, 29(7), 869-876 (2008)
Hyunbum Kim et al.
International journal of molecular sciences, 22(5) (2021-04-04)
The cell-cell/cell-matrix interactions between myoblasts and their extracellular microenvironment have been shown to play a crucial role in the regulation of in vitro myogenic differentiation and in vivo skeletal muscle regeneration. In this study, by harnessing the heparin-mimicking polymer, poly(sodium-4-styrenesulfonate)
Sarah E Pierce et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 15(42), 11244-11255 (2009-09-12)
Isoguanine (2-oxo-6-amino-guanine), a natural but non-standard base, exhibits unique self-association properties compared to its isomer, guanine, and results in formation of different higher order DNA structures. In this work, the higher order structures formed by oligonucleotides containing guanine repeats or

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service