Skip to Content
Merck
All Photos(2)

Key Documents

202762

Sigma-Aldrich

Barium sulfate

99.99% trace metals basis

Synonym(s):

Baryte

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
BaSO4
CAS Number:
Molecular Weight:
233.39
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.22
grade:
for analytical purposes
form:
powder

grade

for analytical purposes

Quality Level

Assay

99.99% trace metals basis

form

powder

SMILES string

[Ba++].[O-]S([O-])(=O)=O

InChI

1S/Ba.H2O4S/c;1-5(2,3)4/h;(H2,1,2,3,4)/q+2;/p-2

InChI key

TZCXTZWJZNENPQ-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Barium sulfate is an insoluble salt commonly used in X-ray imaging as a contrast agent.

Application

Barium sulfate may be used as a catalyst for the conversion of lactic acid to acrylic acid via dehydration.

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Barium sulphate catalyzed dehydration of lactic acid to acrylic acid.
Peng J, et al.
Green Chemistry, 16(1), 108-111 (2014)
Recent advances in the preparation of barium sulfate nanoparticles: a mini-review
Tlek K et al.
Chemical Engineering Journal, 6, 30-30 (2022)
George X Huang et al.
Tissue engineering. Part A, 21(15-16), 2228-2240 (2015-04-30)
Polymeric scaffolds, which release growth factors in a temporally controlled manner, have successfully directed the differentiation of stem cells into monolithic tissues of a single lineage. However, engineering precise boundaries in multilineage functional tissues, such as the juxtaposed cartilaginous and
Tafadzwa P Makarawo et al.
The American surgeon, 79(7), 728-733 (2013-07-03)
Water as an intraluminal negative contrast medium produces improved image quality with reduced artefact. However, rapid absorption of oral water in the bowel relative to speed and timing of image capturing has limited its clinical application. These findings predate advances
Koji Murono et al.
Diseases of the colon and rectum, 57(6), 715-724 (2014-05-09)
Preoperative chemoradiotherapy has been widely used for the prevention of local recurrence of locally advanced rectal cancer, and the effect of chemoradiotherapy is known to be associated with overall survival. We aimed to evaluate the association of the pathologic response

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service