449156
Nickel(II) bromide
anhydrous, powder, ≥99.99% trace metals basis
Synonym(s):
Nickel bromide, Nickel bromide (Br2 Ni), Nickel dibromide, Nickelous bromide
About This Item
Recommended Products
grade
anhydrous
Quality Level
Assay
≥99.99% trace metals basis
form
powder
impurities
≤100.0 ppm Trace Metal Analysis
mp
963 °C (lit.)
density
5.098 g/mL at 25 °C (lit.)
application(s)
battery manufacturing
SMILES string
Br[Ni]Br
InChI
1S/2BrH.Ni/h2*1H;/q;;+2/p-2
InChI key
IPLJNQFXJUCRNH-UHFFFAOYSA-L
Looking for similar products? Visit Product Comparison Guide
accessory
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1B - Muta. 2 - Repr. 1B - Resp. Sens. 1 - Skin Sens. 1 - STOT RE 1 Inhalation
Storage Class Code
6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service