Skip to Content
Merck
All Photos(1)

Documents

678023

Sigma-Aldrich

4,7-Dimethoxy-1,10-phenanthroline

97%

Synonym(s):

4,7-Dimethoxy-1,10-phenanthrolin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H12N2O2
CAS Number:
Molecular Weight:
240.26
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

reaction suitability

reagent type: ligand

mp

197-212 °C

SMILES string

COc1ccnc2c1ccc3c(OC)ccnc23

InChI

1S/C14H12N2O2/c1-17-11-5-7-15-13-9(11)3-4-10-12(18-2)6-8-16-14(10)13/h3-8H,1-2H3

InChI key

ZPGVCQYKXIQWTP-UHFFFAOYSA-N

General description

4,7-Dimethoxy-1,10-phenanthroline is an organic compound that is used as an efficient ligand in organic synthesis. It is used in the N-arylation of imidazole with aryl iodides and bromides in the presence of copper catalysts. Various hindered and functionalized imidazoles and aryl halides were converted using this catalyst.

Application

Ligand for Cu-catalzyed N-arylation of imidazoles

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Artis Klapars et al.
Journal of the American Chemical Society, 124(25), 7421-7428 (2002-06-20)
An experimentally simple and inexpensive catalyst system was developed for the amidation of aryl halides by using 0.2-10 mol % of CuI, 5-20 mol % of a 1,2-diamine ligand, and K(3)PO(4), K(2)CO(3), or Cs(2)CO(3) as base. Catalyst systems based on
Li, C. S.; Dixon, D. D.
Tetrahedron Letters, 45, 4257-4257 (2004)
Ryan A Altman et al.
Organic letters, 9(4), 643-646 (2007-02-09)
With use of Cu-based catalysts, 2- and 4-hydroxypyridines were N-arylated in modest to excellent yields. In the case of 2-hydroxypyridine, the use of 4,7-dimethoxy-1,10-phenanthroline, 3, expanded the scope of previous literature reports to include the use of N-containing heteroaryl halides
Ryan A Altman et al.
Organic letters, 8(13), 2779-2782 (2006-06-16)
[reaction: see text] 4,7-Dimethoxy-1,10-phenanthroline (L) was found to be an efficient ligand for the copper-catalyzed N-arylation of imidazole with aryl iodides and bromides under mild conditions. A variety of hindered and functionalized imidazoles and aryl halides were transformed in good
Ryan A Altman et al.
The Journal of organic chemistry, 72(16), 6190-6199 (2007-07-13)
4,7-Dimethoxy-1,10-phenanthroline (L1c) was found to be an efficient ligand for the copper-catalyzed N-arylation of imidazoles and benzimidazoles with both aryl iodides and bromides under mild conditions. Further optimization of the system has revealed that the addition of poly(ethylene glycol) accelerates

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service