Skip to Content
Merck
All Photos(1)

Documents

334022

Sigma-Aldrich

Cobalt(II) bromide

99%

Synonym(s):

Cobaltous bromide, Dibromocobalt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CoBr2
CAS Number:
Molecular Weight:
218.74
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

Quality Level

Assay

99%

form

powder

mp

678 °C (lit.)

solubility

acetone: soluble(lit.)
ethanol: soluble(lit.)
methanol: soluble(lit.)

density

4.909 g/mL at 25 °C (lit.)

application(s)

battery manufacturing

SMILES string

Br[Co]Br

InChI

1S/2BrH.Co/h2*1H;/q;;+2/p-2

InChI key

BZRRQSJJPUGBAA-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Application


  • Synthesis, crystal structure and thermal properties of dibromidobis (2-methylpyridine N-oxide-κO) cobalt (II): This study provides insights into the crystal structure and thermal properties of a cobalt(II) bromide complex, highlighting its potential applications in materials science (Näther & Jess, 2024).

  • Coordination complexes of isoquinoline with cobalt and nickel halides: The study describes the formation of coordination complexes using cobalt(II) bromide with isoquinoline, which could be used for further exploration in coordination chemistry (Bellerive et al., 2024).

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1B - Muta. 2 - Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A C Blackburn et al.
Acta crystallographica. Section C, Crystal structure communications, 47 ( Pt 2), 282-286 (1991-02-15)
Tetraaquacobalt(II) bromide dihydrate, [Co(H2O)4]Br2 x 2H2O, Mr = 326.83, monoclinic, C2/m, a = 11.0158(12), b = 7.1694(10), c = 6.9111(10) A, beta = 124.761(7)degrees, V = 448.4(1) A3, Z = 2, F(000) = 314, Dx = 2.420 g cm-3, lambda(Mo
Yuefan Wang et al.
Chemical communications (Cambridge, England), 48(66), 8183-8185 (2012-03-24)
A cobalt-TMTU complex, derived from the in situ reduction of CoBr(2) with Zn in the presence of TMTU, can catalyze Pauson-Khand reaction at a balloon pressure of CO, which enables the synthesis of structurally diverse cyclopentenones. This catalytic system works

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service