Skip to Content
Merck
All Photos(1)

Key Documents

446785

Sigma-Aldrich

Perfluorotetradecanoic acid

96%

Synonym(s):

2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11,12,12,13,13,14,14,14-Heptacosafluorotetradecanoic acid, Perfluoro-n-tetradecanoic acid, Perfluoromyristic acid, Perfluorotetradecanecarboxylic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CF3(CF2)12CO2H
CAS Number:
Molecular Weight:
714.11
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

96%

form

solid

bp

270 °C/740 mmHg (lit.)

mp

130-135 °C (lit.)

functional group

carboxylic acid
fluoro

SMILES string

OC(=O)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F

InChI

1S/C14HF27O2/c15-2(16,1(42)43)3(17,18)4(19,20)5(21,22)6(23,24)7(25,26)8(27,28)9(29,30)10(31,32)11(33,34)12(35,36)13(37,38)14(39,40)41/h(H,42,43)

InChI key

RUDINRUXCKIXAJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Perfluorotetradecanoic acid can be used as a reactant:
  • In the metal-free synthesis of organic iodides using N-iodoamides through the Iododecarboxylation method.
  • To synthesize benzimidazole tethered perfluoromacromolecules by reacting with o-phenylene diamine via condensation reaction.
It can also be used as a surfactant in the formation of mixed monolayer films at the air-water interface.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Dam. 1 - Lact. - Repr. 1B - STOT RE 1

Target Organs

Liver

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hessamaddin Younesi Araghi et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 129, 339-344 (2014-04-22)
Photoillumination of mixed films comprised of the photopolymerizable fatty acid 10,12 pentacosadiynoic acid and perfluorotetradecanoic acid deposited onto glass substrates gives rise to the formation of oriented polydiacetylene photopolymer fibers. The degree of polymer fiber orientation was investigated using dual-view
Hyoung Kun Park et al.
Langmuir : the ACS journal of surfaces and colloids, 20(12), 4851-4858 (2005-06-30)
The effect of an acidic surfactant, perfluorotetradecanoic acid (PFTDA), on the gelation of octadecyltrimethoxysilane (ODTMS) monolayers at the air/water interface was investigated using a Langmuir film balance, attenuated total reflection (ATR) infrared spectroscopy, and atomic force microscopy (AFM). The gelation
David Sowah-Kuma et al.
Langmuir : the ACS journal of surfaces and colloids, 34(36), 10673-10683 (2018-08-14)
Synchrotron-based X-ray scattering measurements of phase-separated surfactant monolayers at the air-water interface provide molecular-level structural information about the packing and ordering of film components. In this work, grazing incidence X-ray diffraction (GIXD) and X-ray reflectivity (XR) measurements were used to
Effrosyni Zafeiraki et al.
Chemosphere, 94, 169-176 (2013-12-25)
Perfluorinated compounds (PFCs) are used in food packaging materials as coatings/additives for oil and moisture resistance. In the current study, foodstuff-packaging materials collected from the Greek market, made of paper, paperboard or aluminum foil were analyzed for the determination of
Wei Zhao et al.
Journal of environmental sciences (China), 56, 272-280 (2017-06-03)
Perfluoroalkyl substances (PFASs) are a class of fluorine substituted carboxylic acid, sulfonic acid and alcohol, structurally similar to their corresponding parent compounds. Previous study demonstrated the potential endocrine disruption and reproductive toxicity of perfluorooctane sulfonic acid and perfluorooctanoic acid, two

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service