442690
Lithium formate monohydrate
98%
Synonym(s):
Formic acid lithium salt
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Linear Formula:
HCO2Li · H2O
CAS Number:
Molecular Weight:
69.97
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
Assay:
98%
form:
crystals and lumps
Recommended Products
Quality Level
Assay
98%
form
crystals and lumps
SMILES string
[Li+].[H]O[H].[H]C([O-])=O
InChI
1S/CH2O2.Li.H2O/c2-1-3;;/h1H,(H,2,3);;1H2/q;+1;/p-1
InChI key
RDAKCPVJSPEDON-UHFFFAOYSA-M
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Jin-Sol Shin et al.
Australasian physical & engineering sciences in medicine, 42(3), 811-818 (2019-08-15)
The nondestructive dosimetry achieved with electron paramagnetic resonance (EPR) dosimetry facilitates repetitive recording by the same dosimeter to increase the reliability of data. In precedent studies, solid paraffin was needed as a binder material to make the lithium formate monohydrate
Qianglu Lin et al.
Journal of the American Chemical Society, 139(19), 6644-6653 (2017-04-22)
The use of semiconductor nanocrystal quantum dots (QDs) in optoelectronic devices typically requires postsynthetic chemical surface treatments to enhance electronic coupling between QDs and allow for efficient charge transport in QD films. Despite their importance in solar cells and infrared
Marko Tarvainen et al.
Food chemistry, 173, 1011-1021 (2014-12-04)
Increasing concern of consumers on the safety of synthetic food additives has created high interest in natural preservatives in food industry. Plant extracts produced by supercritical CO2 technology from rosemary (R), oregano (O) and an antimicrobial blend (AB) consisting of
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service