Skip to Content
Merck
All Photos(1)

Key Documents

429961

Sigma-Aldrich

Triethylgermanium hydride

98%

Synonym(s):

Triethylgermane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C2H5)3GeH
CAS Number:
Molecular Weight:
160.83
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.23

Quality Level

Assay

98%

form

liquid

reaction suitability

core: germanium
reagent type: reductant

refractive index

n20/D 1.433 (lit.)

bp

121-122 °C/20 mmHg (lit.)

density

0.994 g/mL at 25 °C (lit.)

SMILES string

CC[GeH](CC)CC

InChI

1S/C6H16Ge/c1-4-7(5-2)6-3/h7H,4-6H2,1-3H3

InChI key

LTLGQZPDSQYFSB-UHFFFAOYSA-N

General description

Triethylgermanium hydride (Ge(C2H5)3H) may be prepared by a three step process:
  • Ge(C2H5)3Br reacted with Na to yield Ge2(C2H5)6
  • Ge2(C2H5)6 treated with Li to form Ge(C2H5)3Li
  • ammonolysis of Ge(C2H5)3Li to give the end product, Ge(C2H5)3H. 1

Application

Reagent undergoes hydrogermylation with alkyl propiolates to give triethylgermylacrylates.

Packaging

Packaged in glass bottles

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

46.4 °F - closed cup

Flash Point(C)

8 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Journal of Organometallic Chemistry, 435, 257-257 (1992)
Theresia Ahrens et al.
Dalton transactions (Cambridge, England : 2003), 45(43), 17495-17507 (2016-11-02)
The reaction of the silyl complex [Rh{Si(OEt)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service