Skip to Content
Merck
All Photos(1)

Key Documents

230197

Sigma-Aldrich

Triethylsilane

Triethylsilane

99%

Synonym(s):

NSC 93579, Triethylhydrosilane, Triethylsilicon hydride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C2H5)3SiH
CAS Number:
Molecular Weight:
116.28
Beilstein:
1098278
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Quality Level

Assay

99%

form

liquid

reaction suitability

reagent type: reductant

refractive index

n20/D 1.412 (lit.)

bp

107-108 °C (lit.)

density

0.728 g/mL at 25 °C (lit.)

SMILES string

CC[SiH](CC)CC

InChI

1S/C6H16Si/c1-4-7(5-2)6-3/h7H,4-6H2,1-3H3

InChI key

AQRLNPVMDITEJU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Catalyst for:
  • Synthesis of a spiro-oxindole blocker of Nav1.7 for the treatment of pain
  • Redox initiated cationic polymerization
  • Beckmann rearrangement of cyclododecanone oxime
  • Regioselective reductive coupling of enones and allenes

Catalyst reactivation after catalyst polymerization of styrene

Studies involving the prediction of organosilicon flash points
Used in a study of the reduction of 2-chromanols; syn-selectivity observed with TES.[1]
Versatile reducing agent

related product

Product No.
Description
Pricing

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

21.2 °F

Flash Point(C)

-6 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kelin Li et al.
Organic letters, 8(21), 4711-4714 (2006-10-06)
[reaction: see text] Reduction of C5-substituted 2-hydroxychromans selectively provides 2,4-cis-chromans using large silane reductants and 2,4-trans-chromans using the smaller silane PhSiH(3). The stereochemical outcome has been rationalized on the basis of a Curtin-Hammett kinetic situation arising from hydride delivery to
Zaihui Zhang et al.
Journal of medicinal chemistry, 56(2), 568-583 (2012-12-19)
Stearoyl-CoA desaturase-1 (SCD1) catalyzes de novo synthesis of monounsaturated fatty acids from saturated fatty acids. Studies have demonstrated that rodents lacking a functional SCD1 gene have an improved metabolic profile, including reduced weight gain, lower triglycerides, and improved insulin response.
Sultan Chowdhury et al.
Bioorganic & medicinal chemistry letters, 21(12), 3676-3681 (2011-05-17)
Starting from the oxindole 2a identified through a high-throughput screening campaign, a series of Na(V)1.7 blockers were developed. Following the elimination of undesirable structural features, preliminary optimization of the oxindole C-3 and N-1 substituents afforded the simplified analogue 9b, which
Emma J Ste Marie et al.
Journal of peptide science : an official publication of the European Peptide Society, 24(11), e3130-e3130 (2018-10-26)
Triisopropylsilane (TIS), a hindered hydrosilane, has long been utilized as a cation scavenger for the removal of amino acid protecting groups during peptide synthesis. However, its ability to actively remove S-protecting groups by serving as a reductant has largely been
Younggyu Kim et al.
Nucleic acids research, 36(19), 6143-6154 (2008-10-01)
We have developed an approach that enables nonradioactive, ultrasensitive (attamole sensitivity) site-specific protein-protein photocrosslinking, and we have applied the approach to the analysis of interactions of alpha-helix 2 (H2) of human TATA-element binding protein (TBP) with general transcription factor TFIIA

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

Active Filters

  1. 1 Ratings-Only Review

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service