Skip to Content
Merck
All Photos(1)

Key Documents

288217

Sigma-Aldrich

Triphenylgermanium chloride

99%

Synonym(s):

Chlorotriphenylgermane, Triphenylchlorogermane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C6H5)3GeCl
CAS Number:
Molecular Weight:
339.40
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

99%

form

solid

bp

285 °C/12 mmHg (lit.)

mp

114-115 °C (lit.)

SMILES string

Cl[Ge](c1ccccc1)(c2ccccc2)c3ccccc3

InChI

1S/C18H15ClGe/c19-20(16-10-4-1-5-11-16,17-12-6-2-7-13-17)18-14-8-3-9-15-18/h1-15H

InChI key

ONQMBYVVEVFYRP-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bruno Pescara et al.
Inorganic chemistry, 58(8), 4802-4811 (2019-03-29)
Germanium (Ge) nanoparticles are gaining increasing interest due to their properties that arise in the quantum confinement regime, such as the development of the band structure with changing size. While promising materials, significant challenges still exist related to the development
Luba Hunakova et al.
Toxicology letters, 279, 16-21 (2017-07-16)
Among a variety of metal containing organic compounds, tin derivatives are enjoying an increasing interest and appear to be very promising as potential drug candidates. We studied eight organometallic derivatives, nuclear retinoid X receptor (RXR) ligands and two germanium containing

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service