Skip to Content
Merck
All Photos(1)

Documents

413658

Sigma-Aldrich

Indium(I) iodide

AnhydroBeads, −10 mesh, 99.999% trace metals basis

Synonym(s):

Indium monoiodide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
InI
CAS Number:
Molecular Weight:
241.72
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

product line

AnhydroBeads

Assay

99.999% trace metals basis

form

beads

reaction suitability

reagent type: catalyst
core: indium

impurities

≤15.0 ppm Trace Metal Analysis

particle size

−10 mesh

SMILES string

[In]I

InChI

1S/HI.In/h1H;/q;+1/p-1

InChI key

FOVZCYAIUZHXGB-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Has been discovered and used as a reagent to propagate some organic reactions, such as cyclization and cleavage of diselenide or disulfide. Can also be used to prepare In-containing complexes.

Legal Information

AnhydroBeads is a trademark of Sigma-Aldrich Co. LLC

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tiwari, P.; et al.
Tetrahedron Letters, 47 (14), 2345-2348 (2006)
Ranu, B.C.; et al.
Tetrahedron Letters, 47 (17), 2859-2861 (2006)
Brindaban C Ranu et al.
The Journal of organic chemistry, 71(1), 423-425 (2006-01-04)
[reaction: see text] Diphenyl diselenide (and disulfide) undergo facile reaction with indium(I) iodide and the corresponding intermediate complex condenses in situ with a variety of substituted vinyl bromides in the presence of a catalytic amount of tetrakis(triphenylphosphine)palladium(0) [Pd(PPh3)4] in THF
Wuping Liao et al.
Inorganic chemistry, 45(10), 3828-3830 (2006-05-09)
A different thermal treatment of identical reactants (EuI2, NaCN, NaN3, and InI) leads to the formation of the three title compounds. In(0.08)Eu4(NCN)3I3 is isotypic with the reported LiEu4(NCN)3I3, Eu8I9(CN)(NCN)3 represents the first mixed cyanide-cyanamide rare-earth compound, and In(0.28)Eu12(NCN)5I(14.91) is characterized
Goeta, A.; et al.
Tetrahedron, 62, 3582-3599 (2006)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service