412422
PAMAM dendrimer, ethylenediamine core, generation 3.0 solution
20 wt. % in methanol
Synonym(s):
Starburst PAMAM-G 3
About This Item
Recommended Products
description
amino surface groups
form
liquid
mol wt
generation 3.0
feature
core type ethylenediamine core (2-carbon core)
concentration
20 wt. % in methanol
no. Surface Groups
32
refractive index
n20/D 1.367
bp
64.7 °C
density
0.863 g/mL at 25 °C
storage temp.
2-8°C
General description
Application
Physical form
Legal Information
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1
Target Organs
Eyes,Central nervous system
Storage Class Code
3 - Flammable liquids
WGK
WGK 3
Flash Point(F)
51.8 °F - closed cup
Flash Point(C)
11 °C - closed cup
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service