Skip to Content
Merck
All Photos(3)

Key Documents

412384

Sigma-Aldrich

PAMAM dendrimer, ethylenediamine core, generation 1.0 solution

20 wt. % in methanol

Synonym(s):

PAMAM dendrimer (G-1)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[NH2(CH2)2NH2]:(G=1);dendri PAMAM(NH2)8
CAS Number:
Molecular Weight:
1429.85
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

description

amino surface groups

form

liquid

mol wt

generation 1.0

feature

core type ethylenediamine core (2-carbon core)

concentration

20 wt. % in methanol

no. Surface Groups

8

refractive index

n20/D 1.362

bp

64.7 °C

density

0.82 g/mL at 25 °C

storage temp.

2-8°C

SMILES string

NCCNC(=O)CCN(CCNC(=O)CCN(CCN(CCC(=O)NCCN(CCC(=O)NCCN)CCC(=O)NCCN)CCC(=O)NCCN(CCC(=O)NCCN)CCC(=O)NCCN)CCC(=O)NCCN(CCC(=O)NCCN)CCC(=O)NCCN)CCC(=O)NCCN

InChI

1S/C62H128N26O12/c63-13-21-71-51(89)1-33-83(34-2-52(90)72-22-14-64)45-29-79-59(97)9-41-87(42-10-60(98)80-30-46-84(35-3-53(91)73-23-15-65)36-4-54(92)74-24-16-66)49-50-88(43-11-61(99)81-31-47-85(37-5-55(93)75-25-17-67)38-6-56(94)76-26-18-68)44-12-62(100)82-32-48-86(39-7-57(95)77-27-19-69)40-8-58(96)78-28-20-70/h1-50,63-70H2,(H,71,89)(H,72,90)(H,73,91)(H,74,92)(H,75,93)(H,76,94)(H,77,95)(H,78,96)(H,79,97)(H,80,98)(H,81,99)(H,82,100)

InChI key

QVWIZHOFYGKROL-UHFFFAOYSA-N

Application

  • Gold-Dendrimer Nanocomposites: Study on the synthesis and morphological characterization of gold-dendrimer nanocomposites using PAMAM dendrimers with ethylenediamine cores, evaluating their potential biomedical applications (Vasile, Serafim, Petre, Giol, 2014).
  • DNA Interaction Modeling: Analysis of complexes formed between DNA and poly(amido amine) dendrimers of different generations, focusing on modeling DNA wrapping and penetration which includes Generation 1.0 dendrimers (Qamhieh, Nylander, Black, Attard, 2014).

Physical form

Contains 8 surface primary amino groups

Legal Information

Manufactured by Dendritech®, Inc.
Dendritech is a registered trademark of Dendritech, Inc.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 1 - STOT SE 3

Target Organs

Eyes,Central nervous system, Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

51.8 °F

Flash Point(C)

11 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service