Recommended Products
description
amino surface groups
feature
core type ethylenediamine core (2-carbon core)
storage temp.
2-8°C
General description
Ethylenediamine-core poly(amidoamine) (PAMAM) dendrimers are highly branched, “tree-like” core-shell nanostructures made up of amide and amine functional subunits.
Application
PAMAM Dendrimer Kit, generations 4-7 can be used to formulate poorly water-soluble drugs to increase water solubility, and therefore enhance the bioavailability of these drugs. It can also be applied to catalysts and organic molecules, where dendrimers function as solubility enhancers and modifiers, allowing to mix otherwise immiscible materials.
Packaging
unit weight includes solvent
Legal Information
Manufactured by Dendritech®, Inc.
Dendritech is a registered trademark of Dendritech, Inc.
Kit Components Also Available Separately
Product No.
Description
SDS
- 412449PAMAM dendrimer, ethylenediamine core, generation 4.0 solution, 10 wt. % in methanol 1 gSDS
- 536709PAMAM dendrimer, ethylenediamine core, generation 5.0 solution, 5 wt. % in methanol 1 gSDS
- 536717PAMAM dendrimer, ethylenediamine core, generation 6.0 solution, 5 wt. % in methanol 1 gSDS
- 536725PAMAM dendrimer, ethylenediamine core, generation 7.0 solution, 5 wt. % in methanol 1 gSDS
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1
Storage Class Code
3 - Flammable liquids
Flash Point(F)
51.8 °F - closed cup
Flash Point(C)
11 °C - closed cup
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
The Analyst, 142(20), 3848-3856 (2017-09-13)
Although consumer exposure to nanomaterials is ever increasing, with potential increased applications in areas such as drug and/or gene delivery, contrast agents and diagnosis, the determination of the cyto- and geno-toxic effects of nanomaterials on human health and the environment
Heck reactions catalyzed by PAMAM-dendrimer encapsulated Pd (0) nanoparticles
Nano Letters, 1(9), 499-501 (2001)
Poly (amidoamine) dendrimers as a pharmaceutical excipient. Are we there yet?
Journal of Pharmaceutical Sciences, 107(1), 75-83 (2018)
PAMAM dendrimer: a pH-controlled nanosponge
Canadian Journal of Chemistry, 95(9), 991-998 (2017)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service