409022
Poly(diallyldimethylammonium chloride) solution
average Mw 200,000-350,000 (medium molecular weight), 20 wt. % in H2O
Synonym(s):
PDADMAC
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
(C8H16ClN)n
CAS Number:
MDL number:
UNSPSC Code:
12162002
NACRES:
NA.23
Recommended Products
form
viscous liquid
Quality Level
mol wt
average Mw 200,000-350,000 (medium molecular weight)
concentration
20 wt. % in H2O
refractive index
n20/D 1.375
viscosity
250-500 cP(25 °C, Brookfield)
density
1.04 g/mL at 25 °C
SMILES string
[Cl-].[N+](CC=C)(CC=C)(C)C
InChI
1S/C8H16N.ClH/c1-5-7-9(3,4)8-6-2;/h5-6H,1-2,7-8H2,3-4H3;1H/q+1;/p-1
InChI key
GQOKIYDTHHZSCJ-UHFFFAOYSA-M
Looking for similar products? Visit Product Comparison Guide
General description
Poly(diallyldimethylammonium chloride) solution (PDADMAC) is a cationic polyelectrolyte prepared by the radical polymerization of diallyldimethylammonium chloride (DADMAC) in presence of a catalyst in the form of an organic peroxide. It is widely utilized as a flocculant.
Application
PDADMAC can be used as an electrolytic solution in a variety of applications such as drug delivery, biomedical systems, and sensors, which include biosensors and chemical sensors.
Storage Class Code
10 - Combustible liquids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Synthesis and use of PolyDADMAC for water purification
Biennial Conference of the Water Institute of Southern Africa, Durban, South Africa, 46(8), 2584-2594 (2002)
Solution properties of poly (diallyldimethylammonium chloride)(PDDA)
Marcelo G, et al.
Polymer, 46(8), 2584-2594 (2005)
pH-responsive carrier system based on carboxylic acid modified mesoporous silica and polyelectrolyte for drug delivery
Yang Q, et al.
Chemistry of Materials, 17(24), 5999-6003 (2005)
Synthesis and solution properties of poly (N-isopropylacrylamide-co-diallyldimethylammonium chloride)
Deng Y and Pelton R
Macromolecules, 28(13), 4617-4621 (1995)
Polyelectrolyte multilayer as matrix for electrochemical deposition of gold clusters: toward super-hydrophobic surface
Zhang X, et al.
Journal of the American Chemical Society, 126(10), 3064-3065 (2004)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service