Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Poly(diallyldimethylammonium chloride) solution
Synonym(s):
PDADMAC
Linear Formula:
(C
8
H
16
ClN)
n
CAS Number:
26062-79-3
Products
Physical Form
Polymer Type
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
409022
Poly(diallyldimethylammonium chloride) solution,
average M
w
200,000-350,000 (medium molecular weight), 20 wt. % in H
2
O
Expand
View Pricing
522376
Poly(diallyldimethylammonium chloride) solution,
average M
w
<100,000 (very low molecular weight), 35 wt. % in H
2
O
Expand
View Pricing
409014
Poly(diallyldimethylammonium chloride) solution,
20 wt. % in H
2
O
Expand
View Pricing
409030
Poly(diallyldimethylammonium chloride) solution,
average M
w
400,000-500,000 (high molecular weight), 20 wt. % in H
2
O
Expand
View Pricing