Skip to Content
Merck
All Photos(1)

Key Documents

561258

Sigma-Aldrich

Poly(4-styrenesulfonic acid) ammonium salt solution

Mw ~200,000, 30 wt. % in H2O

Synonym(s):

PSS

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12162002
NACRES:
NA.23

mol wt

Mw ~200,000

concentration

30 wt. % in H2O

refractive index

n20/D ~1.3961

pH

3-4 (25 °C)

viscosity

~200 cP(~25 °C, Brookfield) (20 rpm)

SMILES string

[S](=O)(=O)([O-])c1ccc(cc1)C=C.[N+H4]

InChI

1S/C8H8O3S.H3N/c1-2-7-3-5-8(6-4-7)12(9,10)11;/h2-6H,1H2,(H,9,10,11);1H3

InChI key

WTSSRZUUXRDFPD-UHFFFAOYSA-N

Application

Effective antistatic coating for specialty paper and synthetic films.

Features and Benefits

Polyelectrolyte
Polyelectrolyte. Good film-forming properties. Highly temperature stable. Shear stable. pH stable

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hongki Kang et al.
ACS nano, 14(9), 11406-11419 (2020-09-05)
Thermoplasmonic effect-based neural stimulation has been suggested as an alternative optical neural stimulation technology without genetic modification. Integration of near-infrared light with plasmonic gold nanoparticles has been demonstrated as a neuromodulation tool on in vitro neuronal network models. In order

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service