Skip to Content
Merck
All Photos(2)

Documents

406651

Sigma-Aldrich

Poly(propylene glycol) bis(2-aminopropyl ether)

average Mn ~230

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH(NH2)CH2[OCH2CH(CH3)]nNH2
CAS Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

form

liquid

Quality Level

mol wt

average Mn ~230

refractive index

n20/D 1.447

density

0.948 g/mL at 25 °C

SMILES string

CC(N)CO.CC(O)CO

InChI

1S/C4H9NO2/c6-3-1-5-2-4(3)7/h3-7H,1-2H2

InChI key

JCZPOYAMKJFOLA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Poly(propylene glycol) bis(2-aminopropyl ether) is a flexible hydrophobic polymer widely used in the preparation of epoxy resins.

Application

Poly(propylene glycol) bis(2-aminopropyl ether) (PEA) can be used as an additive:
  • To passivate the trap states(surface and grain defects) in perovskite solar cells. PEA helps to improve the stability and efficiency of devices.
  • To fabricate nanocomposites for CO2 capture. The ether group of PEA acts as a proton acceptor that helps to improve the thermal stability and amount of CO2 captured.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Skin Corr. 1C

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 2

Flash Point(F)

262.4 °F - closed cup

Flash Point(C)

128 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Modification of hydrogenated Bisphenol A epoxy adhesives using nanomaterials
Il-Nyoung Yoon, et al.
International Journal of Adhesion and Adhesives, 31, 119-125 (2011)
Synthesis and properties of a bio-based epoxy resin from 2, 5-furandicarboxylic acid (FDCA
Jun Deng, et al.
Royal Society of Chemistry Advances, 5, 15930-15939 (2015)
an efficient trap passivator for perovskite solar cells: Poly (propylene glycol) bis (2-aminopropyl ether)
Ningli Chen, et al.
Nano-Micro Letters, 12, 1-13 (2020)
Amine functionalized hierarchical bimodal mesoporous silicas as a promising nanocomposite for highly efficient CO2 capture
Peiyu Zhao, et al.
Journal of CO2 Utilization, 34, 543-557 (2019)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service