Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Poly(propylene glycol) bis(2-aminopropyl ether)
Linear Formula:
CH
3
CH(NH
2
)CH
2
[OCH
2
CH(CH
3
)]
n
NH
2
CAS Number:
9046-10-0
Products
Brand
Sigma-Aldrich (4)
Physical Form
Polymer Composition
Polymer Type
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
406651
Poly(propylene glycol) bis(2-aminopropyl ether),
average M
n
~230
Expand
View Pricing
406686
Poly(propylene glycol) bis(2-aminopropyl ether),
average M
n
~2,000
Expand
View Pricing
406678
Poly(propylene glycol) bis(2-aminopropyl ether),
average M
n
~400
Expand
View Pricing
406694
Poly(propylene glycol) bis(2-aminopropyl ether),
average M
n
~4,000
Expand
View Pricing