Skip to Content
Merck
All Photos(3)

Documents

404446

Sigma-Aldrich

(R,R)-(−)-N,N′-Bis(3,5-di-tert-butylsalicylidene)-1,2-cyclohexanediaminomanganese(III) chloride

greener alternative

Synonym(s):

(R,R)-Jacobsen’s catalyst, Jacobsen’s catalyst

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[[[(CH3)3C]2C6H2-2-(O-)CH=N]2C6H10]MnCl
CAS Number:
Molecular Weight:
635.20
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.22

greener alternative product characteristics

Catalysis
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

mp

330-332 °C (lit.)

greener alternative category

SMILES string

CC(C)(C)c1cc2\C=N\[C@@H]3CCCC[C@H]3\N=C\c4cc(cc(c4O[Mn](Cl)Oc2c(c1)C(C)(C)C)C(C)(C)C)C(C)(C)C

InChI

1S/C36H54N2O2.ClH.Mn/c1-33(2,3)25-17-23(31(39)27(19-25)35(7,8)9)21-37-29-15-13-14-16-30(29)38-22-24-18-26(34(4,5)6)20-28(32(24)40)36(10,11)12;;/h17-22,29-30,39-40H,13-16H2,1-12H3;1H;/q;;+3/p-3/b37-21+,38-22+;;/t29-,30-;;/m1../s1

InChI key

LJVAWOSDJSQANR-OHRASPNLSA-K

General description

(R,R)-(-)-N,N′-Bis(3,5-di-tert-butylsalicylidene)-1,2-cyclohexanediaminomanganese(III) chloride is a metal-salen complex that can catalyze a variety of asymmetric transformations

(R,R)-(−)-N,N′-Bis(3,5-di-tert-butylsalicylidene)-1,2-cyclohexanediaminomanganese(III) chloride is a catalyst used in the epoxidation of arenes.
We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for catalytic efficiency. Click here for more information.

Application

Salen (Mn)-catalyzed asymmetric epoxidation of unfunctionalized olefins. High enantioselectivities and yields are obtained for a variety of substrates, especially cis-alkenes. A few applications include the synthesis of the taxol side chain and cis-1-amino-2-indanol. Jacobsen′s catalyst has also been used in the asymmetric α-hydroxylation of silyl enol ethers. Catalyst for the enantioselective epoxidation of a variety of olefins.

Legal Information

Manufactured under license by Sterling Pharma Solutions Limited, using Jacobsen HKR technology.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Enantioselective arene epoxidation under mild conditions by Jacobsen catalyst: The role of protic solvent and co-catalyst in the activation of hydrogen peroxide
Rocha, Mariana, et al.
Applied Catalysis A: General, 460, 116-123 (2013)
A highly potential analogue of jacobsen catalyst with in-built phase transfer capability in enantioselective epoxidation of nonfunctionalized alkenes
Kureshy, Rukhsana I and Noor-ul, et al.
Chinese Journal of Catalysis, 209, 99-104 (2002)
Journal of the American Chemical Society, 113, 7063-7063 (1991)
Adam, W. et al.
Tetrahedron, 37, 6531-6531 (1996)
Zhang, W. Jacobsen, E.N.
The Journal of Organic Chemistry, 56, 2296-2296 (1991)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service