Skip to Content
Merck
All Photos(1)

Documents

SML0742

Sigma-Aldrich

EUK-8

≥95% (HPLC)

Synonym(s):

(SP-5-13)-Chloro[[2,2′-[1,2-ethanediylbis(nitrilomethylidyne)]bis[phenolato]](2-)-N,N′,O,O′]-manganese, (Salen)manganese(III) chloride, Chloro(bis(salicylidene)ethylenediamine)manganese, Chloro[N,N′-ethylenebis(salicylideneaminato)]manganese, EUK 8, Manganese(salen) chloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H14ClMnN2O2
CAS Number:
Molecular Weight:
356.69
UNSPSC Code:
12352200
NACRES:
NA.77

Assay

≥95% (HPLC)

form

powder

storage condition

desiccated

color

, faint to very dark brown

solubility

H2O: 1 mg/mL, clear

storage temp.

−20°C

InChI

1S/C16H16N2O2.ClH.Mn/c19-15-7-3-1-5-13(15)11-17-9-10-18-12-14-6-2-4-8-16(14)20;;/h1-8,11-12,19-20H,9-10H2;1H;/q;;+3/p-3

InChI key

HBQGFROGWOHBAG-UHFFFAOYSA-K

Biochem/physiol Actions

EUK-8 is a salen manganese complex that exhibit potent antioxidant activities. EUK-8 is a superoxide dismutase (SOD) that inhibits amyloid fibril formation, including islet amyloid polypeptide (IAPP), β-amyloid and lysozyme amyloid aggregation. It appears that EUK-8 disrupts the pre-formed amyloid fibrils. EUK-8 increases the viability of the SK-N-MC cells exposed to preformed IAPP fibrils.

Other Notes

air sensitive

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nirali Pandya et al.
Chirality, 24(12), 1063-1073 (2012-09-13)
We are reporting the synthesis, characterization, and calf thymus DNA binding studies of novel chiral macrocyclic Mn(III) salen complexes S-1, R-1, S-2, and R-2. These chiral complexes showed ability to bind with DNA, where complex S-1 exhibits the highest DNA
Toru Sasaki et al.
Aging cell, 7(4), 459-469 (2008-04-19)
Oxidative stress, an imbalance between endogenous levels of oxygen radicals and antioxidative defense, increases with aging. However, it is not clear which of these two factors is the more critical. To clarify the production of oxygen radicals increases with age
Stefanie J Ophoven et al.
Anticancer research, 30(10), 3967-3979 (2010-11-03)
Salen-manganese complexes, such as EUK-8 and EUK-134 are known as catalase mimetics with superoxide dismutase (SOD) and peroxidase mimetic activity. Here we show that the concentration-dependent use of salen-manganese complexes within the appropriate time window and with parallel control of
Satoru Kawakami et al.
Circulation journal : official journal of the Japanese Circulation Society, 73(11), 2125-2134 (2009-09-15)
Mice lacking manganese-superoxide dismutase (Mn-SOD) activity exhibit the typical pathology of dilated cardiomyopathy (DCM). In the present study, presymptomatic and symptomatic mutant mice were treated with the SOD/catalase mimetic, EUK-8. Presymptomatic heart/muscle-specific Mn-SOD-deficient mice (H/M-Sod2(-/-)) were treated with EUK-8 (30
Can Li et al.
Chemical communications (Cambridge, England), (6)(6), 547-558 (2007-02-01)
This article reviews the recent progress made in asymmetric catalysis in the nanopores of mesoporous materials and periodic mesoporous organosilicas (PMOs). Some examples of chiral catalysts within the nanopores show improved catalytic performance compared to homogeneous catalysts. The factors including

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service