Skip to Content
Merck
All Photos(1)

Documents

404411

Sigma-Aldrich

(R,R)-(−)-N,N′-Bis(3,5-di-tert-butylsalicylidene)-1,2-cyclohexanediamine

98%

Synonym(s):

(R,R)-Jacobsen’s ligand

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[[(CH3)3C]2C6H2-2-(OH)CH=N]2C6H10
CAS Number:
Molecular Weight:
546.83
Beilstein:
5464823
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

solid

optical activity

[α]20/D −315±15°, c = 1 in methylene chloride

mp

205-207 °C (lit.)

SMILES string

CC(C)(C)c1cc(\C=N\[C@@H]2CCCC[C@H]2\N=C\c3cc(cc(c3O)C(C)(C)C)C(C)(C)C)c(O)c(c1)C(C)(C)C

InChI

1S/C36H54N2O2/c1-33(2,3)25-17-23(31(39)27(19-25)35(7,8)9)21-37-29-15-13-14-16-30(29)38-22-24-18-26(34(4,5)6)20-28(32(24)40)36(10,11)12/h17-22,29-30,39-40H,13-16H2,1-12H3/b37-21+,38-22+/t29-,30-/m1/s1

InChI key

FYNXDGNCEBQLGC-LQWPWIHBSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Amylases from Aspergillus oryzae are used to prevent staling in the baking industry, clarify haze from fruit juices and alcoholic beverages, and produce glucose and maltose syrup products.
Versatile ligand for asymmetric catalysis. Ligand used for preparing Jacobsen′s catalyst suitable for enantioselective epoxidation of olefins lacking functional groups.

Lipases are used industrially for the resolution of chiral compounds and the transesterification production of biodiesel.

Biochem/physiol Actions

α-amylase catalyzes the hydrolysis of internal α-1,4-O-glycosidic bonds in starches to release α-anomeric sugars.
Lipases catalyze the hydrolysis of triacylglycerols into glycerol and free fatty acids.

Legal Information

Manufactured under license by Sterling Pharma Solutions Limited, using Jacobsen HKR technology.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Journal of the American Chemical Society, 113, 7063-7063 (1991)
S. Chang et al.
The Journal of Organic Chemistry, 58, 6939-6939 (1993)
E.N. Jacobsen et al.
Journal of the American Chemical Society, 113, 7063-7063 (1991)
L. Deng et al.
The Journal of Organic Chemistry, 57, 4320-4320 (1992)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service