Skip to Content
Merck
All Photos(1)

Key Documents

399027

Sigma-Aldrich

Brucine

anhydrous, 98%

Synonym(s):

10,11-Dimethoxystrychnine, 2,3-Dimethoxystrychnidin-10-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H26N2O4
CAS Number:
Molecular Weight:
394.46
Beilstein:
63046
EC Number:
MDL number:
UNSPSC Code:
12352210
PubChem Substance ID:
NACRES:
NA.22

grade

anhydrous

Assay

98%

optical activity

[α]20/D −118°, c = 2.5 in chloroform

mp

175-178 °C (dec.) (lit.)

functional group

ether

SMILES string

[H][C@@]12CC(=O)N3c4cc(OC)c(OC)cc4[C@]56CC[N@H]7CC(=CCO1)[C@]([H])(C[C@@H]57)[C@]2([H])[C@]36[H]

InChI

1S/C23H26N2O4/c1-27-16-8-14-15(9-17(16)28-2)25-20(26)10-18-21-13-7-19-23(14,22(21)25)4-5-24(19)11-12(13)3-6-29-18/h3,8-9,13,18-19,21-22H,4-7,10-11H2,1-2H3/t13-,18-,19-,21-,22-,23+/m0/s1

InChI key

RRKTZKIUPZVBMF-IBTVXLQLSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Brucine is the major active pharmacological component of Strychnos nux-vomica seeds, which is known for its anti-inflammatory and analgesic activities.

Application

Brucine may be used as chiral resolving agent for the optical resolution of tertiary acetylenic alcohols into enantiomers.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 1 Oral - Acute Tox. 2 Inhalation - Aquatic Chronic 3

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Novel Brucine Gel Transdermal Delivery System Designed for Anti-Inflammatory and Analgesic Activities.
Wu P, et al.
International Journal of Molecular Medicine, 18(4), 757-757 (2017)
Hong-Mei Zhang et al.
Molecular biology reports, 39(4), 4937-4947 (2011-12-14)
The features of brucine (BC) binding to two blood proteins, bovine hemoglobin (BHb), and bovine serum albumin (BSA), were investigated via fluorescence, circular dichroism and UV/Vis absorption spectroscopy. The results revealed that BC caused the fluorescence quenching of blood proteins
S S Agrawal et al.
Life sciences, 89(5-6), 147-158 (2011-06-21)
Brucine (BRU), a natural plant alkaloid is reported to possess cytotoxic and antiproliferative activities. In this study we aimed to investigate its in vitro and in vivo antitumor and antiangiogenic effects. Cell proliferation and viability was assessed using microculture tetrazolium
Mohsen Behpour et al.
Phytochemical analysis : PCA, 23(2), 95-102 (2011-05-28)
Strychnos nux-vomica L. (Loganiaceae), widely used in folk medicine, is grown extensively in southern Asian countries. Its major bioactive constituents are strychnine and brucine, which are frequently used in traditional herbal medicines for treatment of nervous diseases, vomiting and traumatic
Jian-Min Qin et al.
International journal of nanomedicine, 7, 369-379 (2012-02-16)
Hepatocellular carcinoma is difficult to diagnose early, and most patients are already in the late stages of the disease when they are admitted to hospital. The total 5-year survival rate is less than 5%. Recent studies have showed that brucine

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service