Skip to Content
Merck
All Photos(3)

Key Documents

C80407

Sigma-Aldrich

Cinchonidine

96%

Synonym(s):

(−)-Cinchonidine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H22N2O
CAS Number:
Molecular Weight:
294.39
Beilstein:
89690
EC Number:
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

96%

form

powder

optical activity

[α]23/D −109.2°, c = 1.5 in ethanol

mp

204-206 °C (lit.)

SMILES string

O[C@@H]([C@@H]1C[C@@H]2CCN1C[C@@H]2C=C)c3ccnc4ccccc34

InChI

1S/C19H22N2O/c1-2-13-12-21-10-8-14(13)11-18(21)19(22)16-7-9-20-17-6-4-3-5-15(16)17/h2-7,9,13-14,18-19,22H,1,8,10-12H2/t13-,14-,18-,19+/m0/s1

InChI key

KMPWYEUPVWOPIM-KODHJQJWSA-N

Gene Information

human ... CYP2D6(1565)

Looking for similar products? Visit Product Comparison Guide

General description

Cinchonidine is a cinchona alkaloid, which belongs to the orthorhombic crystal system and P212121 space group.

Application

The enantioselective hydrogenation of ethyl pyruvate in the presence of cinchonidine modified platinum/alumina catalyst leads to the enantiomeric excess of ethyl (R)-lactate.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Sens. 1A

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Blaser HU, et al.
Studies in Surface Science and Catalysis, 67, 147-155 (1991)
The alkaloid cinchonidine.
Oleksyn BJ
Acta Crystallographica Section B, Structural Science, Crystal Engineering and Materials, 1832-1834 (1982)
Erik Schmidt et al.
Langmuir : the ACS journal of surfaces and colloids, 23(15), 8087-8093 (2007-06-23)
Cinchona alkaloids are frequently used for chiral modification of supported noble metal catalysts employed in heterogeneous enantioselective hydrogenation. In order to gain molecular insight into the surface processes occurring at the metal/liquid interface, cinchonidine (CD) adsorption on vapor-deposited Rh/Al2O3 films
Atsushi Urakawa et al.
The journal of physical chemistry. A, 112(31), 7250-7255 (2008-07-19)
Conformational space of cinchonidine has been explored by means of ab initio potential and free energy surfaces, and the temperature-induced changes of conformational populations were studied by a combined NOESY-DFT analysis. The DFT-derived potential energy surface investigation identified four new
Angelo Vargas et al.
The journal of physical chemistry. A, 110(3), 1118-1127 (2006-01-20)
The conformational analysis of the synthetic chiral modifier O-phenyl-cinchonidine (PhOCD) used in enantioselective hydrogenations over noble metal catalysts has been performed at a PM3 semiempirical level in vacuum. The minimum energy conformations calculated at the DFT level with a medium-size

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service