Skip to Content
Merck
All Photos(3)

Documents

259225

Sigma-Aldrich

1,5-Diphenylcarbazide

ACS reagent

Synonym(s):

DPC, 1,5-Diphenylcarbohydrazide, sym.-Diphenylcarbazide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5NHNHCONHNHC6H5
CAS Number:
Molecular Weight:
242.28
Beilstein:
752039
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.21

grade

ACS reagent

Quality Level

form

solid

ign. residue

≤0.05%

mp

170-175 °C (lit.)
173-176 °C

solubility

aqueous acetone: passes test

suitability

passes test for sensitivity to chromate

SMILES string

O=C(NNc1ccccc1)NNc2ccccc2

InChI

1S/C13H14N4O/c18-13(16-14-11-7-3-1-4-8-11)17-15-12-9-5-2-6-10-12/h1-10,14-15H,(H2,16,17,18)

InChI key

KSPIHGBHKVISFI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

1,5-Diphenylcarbazide is a colorimetric reagent commonly employed in the determination of trace metals.

Application

1,5-Diphenylcarbazide may be used in the following processes:
  • As a reagent for the quantification of palladium and platinum by spectrophotometry.
  • As a mediator for the functionalization of carbon paste electrodes employed in the estimation of Hg(II) ions by differential pulse voltammetry.
  • Fabrication of gold nanoparticles that can be employed for the detection of chromium (VI) ions via colorimetry.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Spectrophotometric determination of platinum and palladium with 1, 5-diphenylcarbazide
Brajter K, et al.
Fresenius Journal of Analytical Chemistry, 328(1), 598-598 (1987)
Surface-enhanced Raman scattering and DFT investigation of 1, 5-diphenylcarbazide and its metal complexes with Ca (II), Mn (II), Fe (III) and Cu (II).
Szabo L, et al.
Journal of Molecular Structure, 1073, 10-17 (2014)
Kinetic method for the determination of trace amounts of copper (II) in water matrices by its catalytic effect on the oxidation of 1, 5-diphenylcarbazide.
Crespo GA, et al.
Analytica Chimica Acta, 539(1), 317-325 (2005)
Determination of hexavalent chromium by species specific isotope dilution mass spectrometry and ion chromatography-1, 5-diphenylcarbazide spectrophotometry.
Tirez K, et al.
Journal of Analytical Atomic Spectrometry, 18(8), 922-932 (2003)
Photometric and visual detection of Cr (VI) using gold nanoparticles modified with 1, 5-diphenylcarbazide.
Liu, L, et al.
Microchimica Acta, 1-7 (2016)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service