Skip to Content
Merck
All Photos(2)

Key Documents

390089

Sigma-Aldrich

Dehydroisoandrosterone 3-acetate

97%

Synonym(s):

3-β-Acetoxy-5-androsten-17-one, Prasterone acetate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H30O3
CAS Number:
Molecular Weight:
330.46
EC Number:
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

optical activity

[α]20/D +2.4°, c = 2 in ethanol

drug control

regulated under CDSA - not available from Sigma-Aldrich Canada

mp

168-170 °C (lit.)

functional group

ester
ketone

SMILES string

CC(=O)O[C@H]1CC[C@]2(C)[C@H]3CC[C@@]4(C)[C@@H](CCC4=O)[C@@H]3CC=C2C1

InChI

1S/C21H30O3/c1-13(22)24-15-8-10-20(2)14(12-15)4-5-16-17-6-7-19(23)21(17,3)11-9-18(16)20/h4,15-18H,5-12H2,1-3H3/t15-,16-,17-,18-,20-,21-/m0/s1

InChI key

NCMZQTLCXHGLOK-ZKHIMWLXSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Vânia M A Moreira et al.
Steroids, 72(14), 939-948 (2007-09-22)
A series of novel 1H- and 2H-indazole derivatives of the commercially available dehydroepiandrosterone acetate have been synthesized and tested for inhibition of human cytochrome 17alpha-hydroxylase-C(17,20)-lyase (CYP17), androgen receptor (AR) binding affinity, and cytotoxic potential against three prostate cancer (PC) cell
M A Graf et al.
Fertility and sterility, 59(4), 761-767 (1993-04-01)
To elucidate changes in gonadotropin secretion pattern in patients with hyperandrogenemic chronic anovulation of various origins. Hyperandrogenemic patients (n = 32), divided into subgroups according to certain clinical and biochemical criteria, and a control group (n = 9) of regularly
H Shinohara
Experientia, 37(12), 1353-1354 (1981-12-15)
Fragmentation of oocytes was induced in the ovary, via ovulation suppression, by administration of dehydroepiandrosterone acetate (DHA-Ac) to mature cycling rats. The maximal fragmentation ratio, 15.0 +/- 3.2%, was obtained by 10 mg/100 g b.wt/day for 7-day administration of DHA-Ac.
B Leighton et al.
The Journal of nutrition, 117(7), 1287-1290 (1987-07-01)
The present study is a follow-up to the report that dehydroepiandrosterone (DHEA) acetate treatment in rats stimulated metabolic heat production and suppressed serum triglycerides, adiposity and weight gain without affecting food intake. Activities of peroxisomal fatty acyl-coenzyme (CoA) oxidase and
D C Cumming
Cleveland Clinic journal of medicine, 57(2), 161-166 (1990-03-01)
The effects of estrogenic steroid hormones in women are apparent, but the circulating androgen levels are much higher. Compared with serum estrone and estradiol levels, circulating testosterone levels are five to 10 times higher, androstenedione levels 30 times higher, dehydroepiandrosterone

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service