Skip to Content
Merck
All Photos(1)

Key Documents

31579

Supelco

Androsterone

VETRANAL®, analytical standard

Synonym(s):

3α-Hydroxy-5α-androstan-17-one, 5α-Androstan-3α-ol-17-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H30O2
CAS Number:
Molecular Weight:
290.44
Beilstein:
2217626
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

VETRANAL®

shelf life

limited shelf life, expiry date on the label

drug control

regulated under CDSA - not available from Sigma-Aldrich Canada

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

181-184 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

[H][C@@]12CC[C@@]3([H])[C@]4([H])CCC(=O)[C@@]4(C)CC[C@]3([H])[C@@]1(C)CC[C@@H](O)C2

InChI

1S/C19H30O2/c1-18-9-7-13(20)11-12(18)3-4-14-15-5-6-17(21)19(15,2)10-8-16(14)18/h12-16,20H,3-11H2,1-2H3/t12-,13+,14-,15-,16-,18-,19-/m0/s1

InChI key

QGXBDMJGAMFCBF-HLUDHZFRSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Androsterone is a tetracydic compound which forms mono-esters or mono-oxime with one hydroxyl group and one keto group respectively.

Application

Androsterone has been used as working reference standard in identifying natural androgen steroids using liquid chromatography–tandem mass spectrometry (LC–MS–MS) in multiple reaction monitoring (MRM) from bovine and urine samples.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

VETRANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 2 - Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 2

1 of 2

Michael Polet et al.
Drug testing and analysis, 11(11-12), 1656-1665 (2019-04-23)
Steroid detection and identification remain key issues in toxicology, drug testing, medical diagnostics, food safety control, and doping control. In this study, we evaluate the capabilities and usefulness of analyzing non-hydrolyzed sulfated steroids with gas chromatography-mass spectrometry (GC-MS) instead of
Herbert J Tobias et al.
Drug testing and analysis, 10(4), 781-785 (2017-09-30)
High-precision carbon isotope ratio analysis of urinary steroids by gas chromatography-combustion-isotope ratio mass spectrometry (GC-C-IRMS) is the official test to detect illicit doping of synthetic versions of endogenous steroids, such as testosterone. Our group created the first steroid isotopic standards
Amelia Palermo et al.
Analytica chimica acta, 964, 112-122 (2017-03-30)
The urinary steroidal fraction has been extensively explored as non-invasive alternative to monitor pathological conditions as well as to unveil the illicit intake of pseudo-endogenous anabolic steroids in sport. However, the majority of previous approaches involved the a priori selection
Development of a liquid chromatography?tandem mass spectrometry method for the identification of natural androgen steroids and their conjugates in urine samples.
Buiarelli, F., et al.
Analytica Chimica Acta, 526.2, 113-120 (2004)
N Duijvesteijn et al.
Journal of animal science, 90(8), 2465-2475 (2012-03-01)
In the pig industry, male piglets are surgically castrated early in life to prevent boar taint. Boar taint is mainly caused by androstenone and skatole. Androstenone is a pheromone that can be released from the salivary glands when the boar

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service