Skip to Content
Merck
All Photos(1)

Documents

377775

Sigma-Aldrich

3-Bromo-1-(trimethylsilyl)-1-propyne

98%

Synonym(s):

3-(Trimethylsilyl)propargyl bromide, 3-Bromo-1-(trimethylsilyl)-1-propyne

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
BrCH2C≡CSi(CH3)3
CAS Number:
Molecular Weight:
191.14
Beilstein:
2037650
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

liquid

refractive index

n20/D 1.483 (lit.)

bp

44-45 °C/2 mmHg (lit.)

density

1.17 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

C[Si](C)(C)C#CCBr

InChI

1S/C6H11BrSi/c1-8(2,3)6-4-5-7/h5H2,1-3H3

InChI key

GAPRPFRDVCCCHR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

3-Bromo-1-(trimethylsilyl)-1-propyne is reported as a propargylating agent. Selective reaction of 3-bromo-1-(trimethylsilyl)-1-propyne with α-keto ester using indium metal has been studied.

Application

3-Bromo-1-(trimethylsilyl)-1-propyne may be used in the preparation of tetrahydroisoquinoline-3-carboxylic acid derivatives. It may be used as reagent for the alkylation of dianion of β-keto esters at γ-carbon. It may be employed in the preparation of terminal conjugated enynes and allenic alcohols.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

145.4 °F - closed cup

Flash Point(C)

63 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S Kotha et al.
Bioorganic & medicinal chemistry letters, 10(13), 1413-1415 (2000-07-11)
Tetrahydroisoquinoline-3-carboxylic acid derivatives are prepared via a [2 + 2 + 2] cycloaddition reaction as a key step using Wilkinson's and CpCo(CO)2 catalysts.
Liebigs Ann. Chem., 673-673 (1994)
Synthesis, 414-414 (1991)
A convenient allylation of 1, n-dicarbonyl compounds using organoindium reagents.
Lee PH, et al.
Bull. Korean Chem. Soc., 22(12), 2301-2304 (2001)
A Convenient Propargylating Agent. 3-Bromo-1-Trimethylisilyl-1-Propyne.
Miller RB.
Synthetic Communications, 2(5), 267-272 (1972)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service