Skip to Content
Merck
All Photos(1)

Documents

429007

Sigma-Aldrich

1-Propynylmagnesium bromide solution

0.5 M in THF

Synonym(s):

1-Propyn-1-ylmagnesium bromide, Bromo(prop-1-yn-1-yl)magnesium, Bromo-1-propynylmagnesium

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C≡CMgBr
CAS Number:
Molecular Weight:
143.26
Beilstein:
3929146
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

reaction suitability

reaction type: Grignard Reaction

concentration

0.5 M in THF

bp

65-67 °C

density

0.941 g/mL at 25 °C

storage temp.

2-8°C

SMILES string

CC#C[Mg]Br

InChI

1S/C3H3.BrH.Mg/c1-3-2;;/h1H3;1H;/q;;+1/p-1

InChI key

YVJBCTFVENNIOV-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

1-Propynylmagnesiumbromide solution is 1-propynylmagnesium bromide in 0.5M tetrahydrofuran.1-propynylmagnesium bromide solution used as a Grignard reagent in chemical synthesis. This research reagent can be used for chemical and organic transformations. It is used as a regent in the conversion of [11C]CO2 to [11C] amides via Mitsunobu reaction.

Application

Preparation of hydroxy enynes which underwent gold(I)-catalyzed benzannulation leading to tetrahydronaphthalenes.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Carc. 2 - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1B - STOT SE 3 - Water-react 2

Target Organs

Central nervous system, Respiratory system

Supplementary Hazards

Storage Class Code

4.3 - Hazardous materials which set free flammable gases upon contact with water

WGK

WGK 3

Flash Point(F)

30.2 °F - closed cup

Flash Point(C)

-1 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The [3+ 2] cycloaddition route to 5-carbomethoxy-4h-1, 2, 3-triazolo [1, 5-a][1] benzazepines from baylis-hillman acetates of 2-azidobenzaldehydes
Song YS and Lee KJ
Journal of Heterocyclic Chemistry, 43(6), 1721-1724 (2006)
Christiane M Grisé et al.
Organic letters, 8(25), 5905-5908 (2006-12-01)
We report a gold-catalyzed benzannulation of 3-hydroxy-1,5-enynes to generate tetrahydronaphthalenes. This mild process proves to be an effective method to synthesize various metasubstituted aromatic rings in good yields. [reaction: see text]
From [11 C] CO 2 to [11 C] amides: a rapid one-pot synthesis via the Mitsunobu reaction
Bongarzone S, et al.
Chemical Communications (Cambridge, England), 53(38), 5334-5337 (2017)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service