Skip to Content
Merck
All Photos(2)

Key Documents

792055

Sigma-Aldrich

2-Azidoethyl 2-bromoisobutyrate

≥95%

Synonym(s):

2′-Azidoethyl 2-bromoisobutyrate, 2-Azidoethyl 2-bromo-2-methylpropanoate, 2-Azidoethyl-2-bromoisobutyrate, 2-Bromo-2-methyl-propanoic acid 2-azidoethyl ester, Azido-ATRP initiator, Azido-Bib

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H10BrN3O2
CAS Number:
Molecular Weight:
236.07
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

Quality Level

Assay

≥95%

form

liquid

refractive index

n20/D 1.482

density

1.437 g/mL at 25 °C

SMILES string

CC(Br)(C)C(OCCN=[N+]=[N-])=O

InChI

1S/C6H10BrN3O2/c1-6(2,7)5(11)12-4-3-9-10-8/h3-4H2,1-2H3

InChI key

OYSXPOJKJKBNSG-UHFFFAOYSA-N

Application

Bifunctional initiator with a bromoisobutyryl moiety for atom transfer radical polymerization (ATRP) and an azide moiety that can be used in Cu-mediated ligation ("click" chemistry) for biomaterials, carbon nanotubes and graphene sheets.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

General Approach to Individually Dispersed, Highly Soluble, and Conductive Graphene Nanosheets Functionalized by Nitrene Chemistry
Hongkun He, Chao Gao
Chemistry of Materials, 22(17), 5054-5064 (2010)
Click chemistry as a route for the immobilization of well-defined polystyrene onto sheets
Shengtong Sun, Yewen Cao, Jiachun Feng; et al.
Journal of Materials Chemistry, 20(27), 5605-5607 (2010)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service