Skip to Content
Merck
All Photos(1)

Documents

364002

Sigma-Aldrich

(1S,2S)-(−)-1,2-Diphenylethylenediamine

97%

Synonym(s):

(1S,2S)-(−)-1,2-Diamino-1,2-diphenylethane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[C6H5CH(NH2)-]2
CAS Number:
Molecular Weight:
212.29
Beilstein:
3201645
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

optical activity

[α]20/D −102°, c = 1 in ethanol

optical purity

ee: 98% (GLC)

mp

83-85 °C (lit.)

SMILES string

N[C@H]([C@@H](N)c1ccccc1)c2ccccc2

InChI

1S/C14H16N2/c15-13(11-7-3-1-4-8-11)14(16)12-9-5-2-6-10-12/h1-10,13-14H,15-16H2/t13-,14-/m0/s1

InChI key

PONXTPCRRASWKW-KBPBESRZSA-N

Looking for similar products? Visit Product Comparison Guide

General description

1,2-Diphenylethylenediamine is a chiral molecule, generally used as a chiral resolving agent and as a precursor of the chiral auxiliary. It is also used as a chiral solvating agent in NMR study.

Application

Versatile ligand for the formation of metal complexes. Used in the synthesis of chiral tropocoronands which have potential utility in asymmetric catalysis.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

(S,S)-1,2-Diphenylethylenediamine
Chiral Reagents for Asymmetric Synthesis (2003)
Jérôme Long et al.
Science (New York, N.Y.), 367(6478), 671-676 (2020-02-08)
Magnetoelectric (ME) materials combine magnetic and electric polarizabilities in the same phase, offering a basis for developing high-density data storage and spintronic or low-consumption devices owing to the possibility of triggering one property with the other. Such applications require strong
Mukaiyama, T.
Aldrichimica Acta, 29, 59-59 (1996)
Chenier, P.J. et al.
Tetrahedron Letters, 38, 7341-7341 (1997)
M Tellier et al.
Journal of pharmaceutical and biomedical analysis, 9(7), 557-563 (1991-01-01)
A new tyrosine-specific LC assay with pre-column fluorogenic derivatization is described for Tyr-Gly as model peptide. o-Hydroxylation of the tyrosine residue with tyrosinase in the presence of ascorbic acid, followed by oxidation to the corresponding quinone by potassium ferricyanide at

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service