Skip to Content
Merck
All Photos(2)

Key Documents

241091

Sigma-Aldrich

trans-4-Phenyl-3-buten-2-one

≥99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH=CHCOCH3
CAS Number:
Molecular Weight:
146.19
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor pressure

0.01 mmHg ( 25 °C)

Assay

≥99%

form

solid

bp

260-262 °C (lit.)

mp

39-41 °C (lit.)

solubility

alcohol: freely soluble(lit.)
benzene: freely soluble(lit.)
chloroform: freely soluble(lit.)
diethyl ether: freely soluble(lit.)
petroleum ether: very slightly soluble(lit.)
water: very slightly soluble(lit.)

functional group

ketone
phenyl

SMILES string

CC(=O)\C=C\c1ccccc1

InChI

1S/C10H10O/c1-9(11)7-8-10-5-3-2-4-6-10/h2-8H,1H3/b8-7+

InChI key

BWHOZHOGCMHOBV-BQYQJAHWSA-N

General description

trans-4-Phenyl-3-buten-2-one forms trans-4-(4-hydroxyphenyl)-3-buten-2-one (4-OH-PBO) as a metabolite when incubated with liver microsomes of untreated rats in the presence of NADPH.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

>230.0 °F

Flash Point(C)

> 110 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bin Cao et al.
Organic & biomolecular chemistry, 10(6), 1239-1245 (2011-12-20)
An efficient and easy procedure to synthesize the pyridinyl analogues of dibenzylideneacetone (pyr-dba) was developed by the condensation of substituted nicotinaldehyde and acetone in the presence of K(2)CO(3) in toluene-EtOH-H(2)O solvent system. Structurally diverse pyr-dba, including quinolinyl dba, can be
Jiyong Zhang et al.
The Journal of organic chemistry, 71(7), 2918-2921 (2006-03-25)
The first efficient asymmetric synthesis of obolactone 1 has been accomplished in 11 steps and with a 15% overall yield in which Brown's enantioselective allylation reactions and ring-closing metathesis reaction are key steps.
Bowon Kwon et al.
Journal of economic entomology, 101(1), 36-41 (2008-03-12)
Benzylideneacetone (BZA) is a metabolite of gram-negative entomopathogenic bacterium Xenorhabdus nematophila, and it acts as an enzyme inhibitor against phospholipase A2 (PLA2). PLA2 catalyzes a committed biosynthetic step of eicosanoids, which mediate insect immune reactions to infection by microbial pathogens.
Samyeol Seo et al.
Applied and environmental microbiology, 78(11), 3816-3823 (2012-03-27)
The entomopathogenic bacteria Xenorhabdus nematophila and Photorhabdus temperata subsp. temperata suppress insect immune responses by inhibiting the catalytic activity of phospholipase A(2) (PLA(2)), which results in preventing biosynthesis of immune-mediating eicosanoids. This study identified PLA(2) inhibitors derived from culture broths
Lisa Dalla Via et al.
European journal of medicinal chemistry, 44(7), 2854-2861 (2009-01-22)
A series of conjugates of alpha,beta-unsaturated ketone systems, phenyl-butenone and diaryl-propenones (chalcones), with the tricyclic planar pyrroloquinoline nucleus were synthesised and evaluated for their anticancer properties. The aim was to target DNA by butenone and chalcones, and determine the occurrence

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service