Skip to Content
Merck
All Photos(1)

Documents

359904

Sigma-Aldrich

3,3-Dimethylbutyraldehyde

95%

Synonym(s):

3,3-Dimethylbutanal, tert-Butylacetaldehyde

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3CCH2CHO
CAS Number:
Molecular Weight:
100.16
Beilstein:
1735829
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

95%

form

liquid

refractive index

n20/D 1.397 (lit.)

bp

104-106 °C (lit.)

density

0.798 g/mL at 25 °C (lit.)

SMILES string

CC(C)(C)CC=O

InChI

1S/C6H12O/c1-6(2,3)4-5-7/h5H,4H2,1-3H3

InChI key

LTNUSYNQZJZUSY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

55.4 °F - closed cup

Flash Point(C)

13 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

LIN H and LIN J.
Fine and Specialty Chemicals / Jing Xi Yu Zhuan Yong Hua Xue Pin, 15, 005-005 (2005)
A convenient synthesis of 3, 3-dimethylbutyraldehyde.
Guo Z, et al.
Synthetic Communications, 31(21), 3395-3399 (2001)
Synlett, 2548-2548 (2006)
Preparation of 3, 3-Dimethylbutyraldehyde [J].
Guo-qin CHEN, et al.
Journal of Xiamen University (Natural Science), 4, 021-021 (2009)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service