Skip to Content
Merck
All Photos(2)

Key Documents

240788

Sigma-Aldrich

Isobutyraldehyde

≥99%

Synonym(s):

2-Methylpropanal, 2-Methylpropionaldehyde

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHCHO
CAS Number:
Molecular Weight:
72.11
Beilstein:
605330
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

vapor density

2.5 (vs air)

vapor pressure

66 mmHg ( 4.4 °C)

Assay

≥99%

form

liquid

autoignition temp.

384 °F

expl. lim.

10 %, 25 °F
2 %, 32 °F

refractive index

n20/D 1.374 (lit.)

bp

63 °C (lit.)

mp

−65 °C (lit.)

solubility

water: soluble 11g/100mL at 20 °C(lit.)
acetone: miscible(lit.)
benzene: miscible(lit.)
carbon disulfide: miscible(lit.)
chloroform: miscible(lit.)
diethyl ether: miscible(lit.)
ethanol: miscible(lit.)
toluene: miscible(lit.)

density

0.79 g/mL at 25 °C (lit.)

functional group

aldehyde

storage temp.

2-8°C

SMILES string

[H]C(=O)C(C)C

InChI

1S/C4H8O/c1-4(2)3-5/h3-4H,1-2H3

InChI key

AMIMRNSIRUDHCM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Isobutyraldehyde undergoes asymmetric Michael addition reaction with β-nitroalkenes to give quaternary carbon-containing nitroalkanes[1].

Application

Isobutyraldehyde (2-methylpropionaldehyde) was used in the microbial synthesis of isobutanol (2-methylpropan-1-ol)[2].

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

-11.2 °F - closed cup

Flash Point(C)

-24 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xiang Liu et al.
Journal of biotechnology, 164(2), 188-195 (2012-09-15)
We have determined the X-ray crystal structures of the NADH-dependent alcohol dehydrogenase LlAdhA from Lactococcus lactis and its laboratory-evolved variant LlAdhA(RE1) at 1.9Å and 2.5Å resolution, respectively. LlAdhA(RE1), which contains three amino acid mutations (Y50F, I212T, and L264V), was engineered
P Bustillo Trueba et al.
Journal of chromatography. A, 1604, 460467-460467 (2019-08-28)
This paper describes the method validation for the simultaneous determination of seven cysteinylated aldehydes, i.e. 2-substituted 1,3-thiazolidines-4-carboxylic acids, using ultra-high-performance liquid chromatography-mass spectrometry (UHPLC-MS). Authentic reference compounds were first synthesized for identification and quantification purposes. Moreover, nuclear magnetic resonance (1H
Weronika Filipowska et al.
Journal of chromatography. A, 1612, 460647-460647 (2019-11-27)
Aldehydes originating from malt play an important role in beer flavour deterioration. In order to better understand the influence of malting process on beer staling, it is necessary to acquire a reliable analytical methodology for determination of beer staling aldehydes
Rudy Coquet et al.
Physical chemistry chemical physics : PCCP, 9(45), 6040-6046 (2007-11-16)
The formation and catalytic behavior of active structures in a SiO(2)-supported unsaturated Ru complex catalyst for alkene epoxidation were studied by means of hybrid density functional theory (DFT) calculations. An energy-gaining route for the catalyst activation was found to allow
Christine Counet et al.
Journal of agricultural and food chemistry, 50(8), 2385-2391 (2002-04-04)
After vacuum distillation and liquid-liquid extraction, the volatile fractions of dark chocolates were analyzed by gas chromatography-olfactometry and gas chromatography-mass spectrometry. Aroma extract dilution analysis revealed the presence of 33 potent odorants in the neutral/basic fraction. Three of these had

Questions

1–2 of 2 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service