Skip to Content
Merck
All Photos(2)

Key Documents

261203

Sigma-Aldrich

Samarium

−40 mesh, 99% trace rare earth metals basis

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Sm
CAS Number:
Molecular Weight:
150.36
EC Number:
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.23
Pricing and availability is not currently available.

Quality Level

Assay

99% trace rare earth metals basis

form

(powder or filings)

reaction suitability

reagent type: catalyst
core: samarium

resistivity

91.4 μΩ-cm, 0°C

particle size

−40 mesh

bp

1794 °C (lit.)

mp

1074 °C (lit.)

density

7.47 g/mL at 25 °C (lit.)

SMILES string

[Sm]

InChI

1S/Sm

InChI key

KZUNJOHGWZRPMI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Arylsulfonyl chlorides and arylsulfinates are reduced by a combination of samarium and titanium(IV) chloride to make diaryl disulfides.[1]

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Sol. 2 - Water-react 3

Storage Class Code

4.3 - Hazardous materials which set free flammable gases upon contact with water

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wang, J. Zhang, Y.
Synthetic Communications, 26, 135-135 (1995)
Yi Lu et al.
Optics express, 21(2), 1997-2002 (2013-02-08)
We demonstrate an all-fiber passively Q-switched Yb-doped laser using a piece of Sm-doped fiber as a saturable absorber. The laser was pumped by two 25W, 975 nm fiber coupled diodes and Q-switching was initiated when the ASE generated in the
M Saif et al.
Chemosphere, 90(2), 840-847 (2012-11-06)
Highly active samarium doped zinc oxide self-cleaning and biocidal surfaces (x mol% Sm(3+)/ZnO where x=0, 1, 2 and 4 mol%) with crystalline porous structures were synthesized by hydrothermal method. Sm(3+)/ZnO thin films were characterized by X-ray diffraction (XRD), transmission electron
M Majumder et al.
Journal of physics. Condensed matter : an Institute of Physics journal, 25(2), 025701-025701 (2012-12-05)
We report resistivity, magnetization and (19)F NMR results in a polycrystalline sample of SmFeAsO(0.86)F(0.14). The resistivity and magnetization data show a sharp drop at 48 K indicating a superconducting transition. The nuclear spin-lattice rate (1/T(1)) and spin-spin relaxation rate (1/T(2))
Hua Yang et al.
Inorganic chemistry, 52(3), 1275-1284 (2013-01-25)
Sm@C(2v)(3)-C(80) has been separated from the carbon soot produced by electrical arc vaporization of graphite rods doped with Sm(2)O(3) and purified. Its structure has been determined by single crystal X-ray diffraction using cocrystals obtained from either Ni(II)(octaethylporphyrin) (Ni(II)(OEP)) to form

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service