Skip to Content
Merck
All Photos(3)

Key Documents

331309

Sigma-Aldrich

Yttrium(III) nitrate tetrahydrate

99.99% trace metals basis

Synonym(s):

Yttrium nitrate tetrahydrate, Yttrium trinitrate tetrahydrate, Yttrium(3+) tetrahydrate trinitronate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Y(NO3)3 · 4H2O
CAS Number:
Molecular Weight:
346.98
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
form:
powder and chunks

Quality Level

Assay

99.99% trace metals basis

form

powder and chunks

impurities

≤150.0 ppm Trace Rare Earth Analysis

density

2.68 g/mL at 25 °C (lit.)

SMILES string

[Y+3].[H]O[H].[H]O[H].[H]O[H].[H]O[H].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O

InChI

1S/3NO3.4H2O.Y/c3*2-1(3)4;;;;;/h;;;4*1H2;/q3*-1;;;;;+3

InChI key

XRKAJAPEZSHSLG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Source of yttrium used to make yttrium-based surfactant mesophases which are promising as adsorbing agents as well as for optically functional species.

Pictograms

Flame over circleExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Ox. Sol. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mitsunori Yada et al.
Inorganic chemistry, 37(25), 6470-6475 (2001-10-24)
The layered and hexagonal yttrium-based surfactant mesophases templated by anionic surfactant (C(n)()H(2)(n)()(+1)OSO(3)(-) and C(n)()H(2)(n)()(+1)SO(3)(-)) assemblies were synthesized by the homogeneous precipitation method using urea. The layered mesophase is formed of a layered but curving or bending microstructure and transformed into

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service