Skip to Content
Merck
All Photos(1)

Documents

326976

Sigma-Aldrich

Silver

foil, thickness 2.0 mm, 99.9% trace metals basis

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Ag
CAS Number:
Molecular Weight:
107.87
EC Number:
MDL number:
UNSPSC Code:
12141740
PubChem Substance ID:
NACRES:
NA.23

Quality Level

Assay

99.9% trace metals basis

form

foil

resistivity

1.59 μΩ-cm, 20°C

thickness

2.0 mm

bp

2212 °C (lit.)

mp

960 °C (lit.)

density

10.49 g/cm3 (lit.)

SMILES string

[Ag]

InChI

1S/Ag

InChI key

BQCADISMDOOEFD-UHFFFAOYSA-N

General description

Silver is a noble metal, extensively used in SERS, photocatalysis and solar cells. The surface of silver can be functionalized to attain specific properties such as biocompatibility and vapor selectivity of sensors.3 Iodized silver foils and thin films find potential use as SERS-active metal substrates.1 Cu substrates laminated with Ag foils, have compatible coefficient of thermal expansion (CTE), to be used for electronic packaging. Porous ZnO nanoplates deposited on silver foil with tunable hydrophobicity may be fabricated.3

Quantity

25 × 25 mm (approximately 13.2 g)

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 2

1 of 2

Direct laminating of silve foils on copper substrate.
Wang PJ, et al.
Journal of Materials Science: Materials in Electronics, 19, 1097-1099 (2008)
Christopher G Jesudason
International journal of molecular sciences, 10(5), 2203-2251 (2009-07-01)
This review is variously a presentation, reflection, synthesis and report with reference to more recent developments of an article - in a journal which has ceased publication - entitled "Some Electrode Theorems with Experimental Corroboration, Inclusive of the Ag/AgCl System"
R Nirmala et al.
Journal of nanoscience and nanotechnology, 13(7), 4686-4693 (2013-08-02)
We report on the preparation and characterization of polyurethane (PU) nanofibers containing silver (Ag) nanoparticles were synthesized by using electrospinning. Two different approaches were adopted to incorporate the Ag nanoparticles in to PU nanofibers. In the first approach, a homogeneous
Rui Wang et al.
Journal of nanoscience and nanotechnology, 13(6), 3851-3854 (2013-07-19)
The present studies reveal that silver nanoparticles (AgNPs) can induce apoptosis and enhance radio-sensitivity on cancer cells. In this paper, we mainly investigated the effect of AgNPs on rat glioma C6 cells upon the combination treatment of hyperthermia treatment (HTT).
Sa Ram Lee et al.
Journal of biomedical nanotechnology, 9(7), 1241-1244 (2013-08-06)
We demonstrate simultaneous detection of surface-enhanced Raman scattering (SERS) and fluorescence signals from a silver microbead. For the dual signal generation, silver microbeads with a diameter of 15 microm were functionalized with benzenethiol (BT) as a Raman tag and a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service