Merck
  • Enhancement effect of cytotoxicity response of silver nanoparticles combined with thermotherapy on C6 rat glioma cells.
Enhancement effect of cytotoxicity response of silver nanoparticles combined with thermotherapy on C6 rat glioma cells.
Journal of nanoscience and nanotechnology (2013-07-19)
Rui Wang, Chunmei Chen, Weizhong Yang, Songsheng Shi, Chunhua Wang, Jing Chen
ABSTRACT
The present studies reveal that silver nanoparticles (AgNPs) can induce apoptosis and enhance radio-sensitivity on cancer cells. In this paper, we mainly investigated the effect of AgNPs on rat glioma C6 cells upon the combination treatment of hyperthermia treatment (HTT). AgNPs were synthesized by a polyol process and the mean size was 15 nm. The particles showed dose-dependent cytotoxicity on C6 cells from the experimental data. Besides, we found that heating cells could enhance the contents of cell uptake of AgNPs. From the survival curves, AgNPs showed the ability to enhance thermo-sensitivity on C6 cells. Our results revealed that AgNPs could have a potential application in enhancing effect on HTT induced killing of glioma cells.
Sigma-Aldrich
Silver nanowires, diam. × L 50 nm × 6 μm, 0.5% (isopropyl alcohol suspension)
Sigma-Aldrich
Silver, nanopowder, <150 nm particle size, 99% trace metals basis
Sigma-Aldrich
Silver, foil, thickness 1.0 mm, 99.99% trace metals basis
Sigma-Aldrich
Silver, shot, 1-3 mm, ≥99.99% trace metals basis
Sigma-Aldrich
Silver, powder, <250 μm, 99.99% trace metals basis
Sigma-Aldrich
Silver, wool, diam. 0.05 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Silver, foil, thickness 0.1 mm, 99.9% trace metals basis
Sigma-Aldrich
Silver, colloidal, 65-75% Ag basis
Sigma-Aldrich
Silver, foil, thickness 2.0 mm, 99.9% trace metals basis
Sigma-Aldrich
Silver, wire, diam. 0.5 mm, ≥99.99% trace metals basis
Sigma-Aldrich
Silver, wire, diam. 1.0 mm, ≥99.99% trace metals basis
Sigma-Aldrich
Silver, wire, diam. 1.0 mm, 99.9% trace metals basis
Sigma-Aldrich
Silver, wire, diam. 0.127 mm, 99.99% trace metals basis
Sigma-Aldrich
Silver, wire, diam. 0.1 mm, 99.9% trace metals basis
Sigma-Aldrich
Silver, conductive paste
Sigma-Aldrich
Silver nanowires, diam. × L 100 nm × 6 μm, 0.5% (isopropyl alcohol suspension)
Sigma-Aldrich
Silver, nanopowder, <100 nm particle size, contains PVP as dispersant, 99.5% trace metals basis
Sigma-Aldrich
Silver, wire, diam. 2.0 mm, 99.9% trace metals basis
Sigma-Aldrich
Silver, wire, diam. 0.25 mm, 99.9% trace metals basis
Sigma-Aldrich
Silver, foil, thickness 0.25 mm, 99.9% trace metals basis
Sigma-Aldrich
Silver, foil, thickness 0.025 mm, 99.9% trace metals basis
Sigma-Aldrich
Silver, powder, 2-3.5 μm, ≥99.9% trace metals basis
Sigma-Aldrich
Silver, wire, diam. 0.25 mm, ≥99.99% trace metals basis
Sigma-Aldrich
Silver, powder, 5-8 μm, ≥99.9% trace metals basis
Sigma-Aldrich
Silver, wire, diam. 2.0 mm, ≥99.99% trace metals basis
Sigma-Aldrich
Silver, granular, >250 μm, 99.99% trace metals basis
Sigma-Aldrich
Silver, evaporation slug, diam. × L 0.6 cm × 1.2 cm, 99.99% trace metals basis
Sigma-Aldrich
Silver, wire, diam. 1.5 mm, ≥99.99% trace metals basis
Sigma-Aldrich
Silver, wire, diam. 0.5 mm, 99.9% trace metals basis
Sigma-Aldrich
Silver, flakes, 10 μm, ≥99.9% trace metals basis