Skip to Content
Merck
All Photos(2)

Key Documents

325805

Sigma-Aldrich

Neodymium(III) acetate hydrate

99.9%

Synonym(s):

Neodymium(III) acetic acid salt hydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3CO2)3Nd · xH2O
CAS Number:
Molecular Weight:
321.37 (anhydrous basis)
EC Number:
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

99.9%

reaction suitability

core: neodymium
reagent type: catalyst

SMILES string

O.CC(=O)O[Nd](OC(C)=O)OC(C)=O

InChI

1S/3C2H4O2.Nd.H2O/c3*1-2(3)4;;/h3*1H3,(H,3,4);;1H2/q;;;+3;/p-3

InChI key

GMQPBTKWMZBSCT-UHFFFAOYSA-K

Application

Neodymium(III) acetate hydrate is used as a doping agent in the synthesis of:
  • Neodymium-doped silica-glasses.
  • Neodymium doped yttrium silicate nanophosphors.(2)

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Microstructural and optical properties of SiO2 glasses doped with ZnSe quantum dots and Nd3+ ions}
El Boukhari T, et al.
Physica B: Condensed Matter, 509, 41-45 (2017)
Liangliang Liang et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 22(31), 10801-10807 (2016-06-02)
A crystal design strategy is described that generates hexagonal-phased NaYF4 :Nd/Yb@NaYF4 :Yb/Tm luminescent nanocrystals with the ability to emit light at 803 nm when illuminated at 745 nm. This is accomplished by taking advantage of the large absorption cross-section of Nd(3+) between
Daifeng Li et al.
Advanced science (Weinheim, Baden-Wurttemberg, Germany), 6(23), 1902042-1902042 (2019-12-14)
Recently, various second near-infrared window (NIR-II, 1000-1700 nm) fluorophores have been synthesized for in vivo imaging with nonradiation, high resolution, and low autofluorescence. However, most of the NIR-II fluorophores, especially inorganic nanoprobes, are mainly retained in the reticuloendothelial system (RES)
Xiao Wu et al.
ACS applied materials & interfaces, 11(1), 385-393 (2018-12-18)
The efficacy of dynamic therapy for solid tumors suffers daunting challenges induced by tumor hypoxia. Herein, we report a biocompatible nanosystem containing Fe(OH)3-modified upconversion nanoparticles (UCNPs) for promoting synergetic chemo- and photodynamic therapy with the modulation of tumor hypoxia. In
Syue-Liang Lin et al.
Nanomaterials (Basel, Switzerland), 10(10) (2020-10-21)
Several robust titania (TiO2) coated core/multishell trivalent lanthanide (Ln) upconversion nanoparticles (UCNPs) hybrid architecture designs have been reported for use in photodynamic therapy (PDT) against cancer, utilizing the near-infrared (NIR) excited energy down-shifting and up-conversion chain of Nd3+ (λ793-808 nm)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service