Skip to Content
Merck

309877

Sigma-Aldrich

4-Bromo-2-nitrophenol

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
BrC6H3(NO2)OH
CAS Number:
Molecular Weight:
218.00
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Pricing and availability is not currently available.

Quality Level

Assay

98%

mp

90-94 °C (lit.)

functional group

bromo
nitro

SMILES string

Oc1ccc(Br)cc1[N+]([O-])=O

InChI

1S/C6H4BrNO3/c7-4-1-2-6(9)5(3-4)8(10)11/h1-3,9H

InChI key

CUTFAPGINUFNQM-UHFFFAOYSA-N

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
532878283541392383
Quality Level

200

Quality Level

100

Quality Level

100

Quality Level

100

mp

90-94 °C (lit.)

mp

38-44 °C (lit.)

mp

110-112 °C (lit.)

mp

211-215 °C (lit.)

functional group

bromo

functional group

bromo, ester

functional group

isothiocyanate, nitro

functional group

bromo, carboxylic acid, fluoro

General description

The Suzuki coupling of 4-bromo-2-nitrophenol with boronic acid was studied[1].

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Adwoa Akwabi-Ameyaw et al.
Bioorganic & medicinal chemistry letters, 19(16), 4733-4739 (2009-07-10)
Two series of conformationally constrained analogs of the FXR agonist GW 4064 1 were prepared. Replacement of the metabolically labile stilbene with either benzothiophene or naphthalene rings led to the identification of potent full agonists 2a and 2g.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service