Skip to Content
Merck
All Photos(1)

Key Documents

532878

Sigma-Aldrich

Methyl 4-bromo-3-methylbenzoate

95%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
BrC6H3(CH3)CO2CH3
CAS Number:
Molecular Weight:
229.07
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

mp

38-44 °C (lit.)

SMILES string

COC(=O)c1ccc(Br)c(C)c1

InChI

1S/C9H9BrO2/c1-6-5-7(9(11)12-2)3-4-8(6)10/h3-5H,1-2H3

InChI key

GTZTYNPAPQKIIR-UHFFFAOYSA-N

General description

Methyl 4-bromo-3-methylbenzoate, an aromatic ester, undergoes Suzuki coupling with 2-chloroboronic acid to afford the corresponding biaryl.

Application

Methyl 4-bromo-3-methylbenzoate may be used in the preparation of:
  • 4-bromo-3-vinylbenzoic acid
  • 4-bromo-3-(methylphenyl)methanol
  • methyl (E)-4-bromo-3-[(phenylmethoxyimino)methyl]benzoate

It may be used as a starting material in the total synthesis of (-)-martinellic acid.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis and properties of mesogen-jacketed liquid crystalline polymers with asymmetry mesogenic core.
Chen S, et al.
Macromolecules, 40(16), 5718-5725 (2007)
Atsushi Shirai et al.
The Journal of organic chemistry, 73(12), 4464-4475 (2008-05-14)
The asymmetric total synthesis of martinellic acid, the first pyrrolo[3,2-c]quinoline alkaloid found in nature, is described. Three key steps in our synthesis of (-)-martinellic acid are the Bu(3)SnH-promoted radical addition-cyclization-elimination (RACE) reaction of an oxime ether with an alpha,beta-unsaturated ester
Sheel C Dodani et al.
Proceedings of the National Academy of Sciences of the United States of America, 111(46), 16280-16285 (2014-11-08)
For reasons that remain insufficiently understood, the brain requires among the highest levels of metals in the body for normal function. The traditional paradigm for this organ and others is that fluxes of alkali and alkaline earth metals are required
Steven Harper et al.
Journal of medicinal chemistry, 48(5), 1314-1317 (2005-03-04)
Allosteric inhibition of the hepatitis C virus (HCV) NS5B RNA-dependent RNA polymerase enzyme has recently emerged as a viable strategy toward blocking replication of viral RNA in cell-based systems. We report here a novel class of allosteric inhibitor of NS5B
Hengye Li et al.
Analytica chimica acta, 985, 91-100 (2017-09-03)
4-Carboxy-benzoboroxole was designed and synthesized. It was then combined with the modification effect of polyethyleneimine (PEI) for the preparation of boronate affinity silica stationary phase. The stationary phase showed improved binding strength with dissociation constant (K

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service