Skip to Content
Merck
All Photos(3)

Key Documents

303666

Sigma-Aldrich

2-Butynoic acid

98%

Synonym(s):

Tetrolic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C≡CCO2H
CAS Number:
Molecular Weight:
84.07
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

form

solid

mp

78-80 °C (lit.)

functional group

carboxylic acid

SMILES string

CC#CC(O)=O

InChI

1S/C4H4O2/c1-2-3-4(5)6/h1H3,(H,5,6)

InChI key

LUEHNHVFDCZTGL-UHFFFAOYSA-N

General description

The rotational spectrum of 2-butynoic acid was measured by pulsed supersonic-jet Fourier transform microwave spectroscopy.

Application

2-Butynoic acid was employed as synthon in a variety of reactions, including cycloacylation of phenols to flavones and chromones, and cyclization to γ-butyrolactones. It was also used in synthesis of Z-trisubstituted olefins via γ-alkylation.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The Journal of Organic Chemistry, 55, 4349-4349 (1990)
I A Mjör et al.
Journal of dental research, 61(8), 967-972 (1982-08-01)
Adhesive bonding of resins to dentin surfaces requires the removal of the layer of debris caused by the cutting. Certain isotonic acidic solutions can do this rapidly. Five solutions were evaluated using cell cultures and pulp studies in monkeys. At
The Journal of Organic Chemistry, 56, 5120-5120 (1991)
Almost free methyl top internal rotation: Rotational spectrum of 2-butynoic acid.
Ilyushin V, et al.
Journal of Molecular Spectroscopy, 267(1), 186-190 (2011)
gamma.-Alkylation of 2-butynoic acid. Route to controlled prenol homologation.
Pitzele BS, et al.
The Journal of Organic Chemistry, 40(2), 269-270 (1975)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service