Skip to Content
Merck
All Photos(3)

Documents

P51400

Sigma-Aldrich

Propiolic acid

95%

Synonym(s):

Acetylenecarboxylic acid, Propynoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HC≡CCOOH
CAS Number:
Molecular Weight:
70.05
Beilstein:
878176
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

95%

impurities

≤6.0% acetic acid

refractive index

n20/D 1.431 (lit.)

bp

102 °C/200 mmHg (lit.)

mp

16-18 °C (lit.)

density

1.138 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

OC(=O)C#C

InChI

1S/C3H2O2/c1-2-3(4)5/h1H,(H,4,5)

InChI key

UORVCLMRJXCDCP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Reagent employed in the synthesis of transition metal complexes, haloalkyl propiolates, and halopropenoates.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Dermal - Acute Tox. 3 Oral - Eye Dam. 1 - Flam. Liq. 3 - Skin Corr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

136.4 °F - closed cup

Flash Point(C)

58 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tetrahedron, 49, 4229-4229 (1993)
The Journal of Organic Chemistry, 57, 709-709 (1992)
Journal of Organometallic Chemistry, 456, 271-271 (1993)
Tina N Grant et al.
Organic letters, 10(18), 3985-3988 (2008-08-19)
Readily available five- and six-membered lactones and N-sulfonyllactams undergo efficient addition of t-butyl propiolate, and the resulting adducts undergo cycloisomerization to six- and seven-membered cyclic ethers or amines in the presence of pyridinium acetate. The ring expansion process occurs in
Jihye Park et al.
The Journal of organic chemistry, 76(7), 2214-2219 (2011-03-05)
One-pot synthesis of symmetric 1,4-disubstituted 1,3-diynes from iodoarenes and propiolic acid via Sonogashira reaction followed by Pd-catalyzed decarboxylative homocoupling is developed in high yields. Also, this system allows the one-pot synthesis of unsymmetric 1,4-disubstituted 1,3-diynes by cross-coupling of two different

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service