Skip to Content
Merck
All Photos(1)

Key Documents

302767

Sigma-Aldrich

(S)-(−)-1-Methyl-2-pyrrolidinemethanol

96%

Synonym(s):

N-Methyl-L-prolinol, (S)-2-Hydroxymethyl-1-methylpyrrolidine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H13NO
CAS Number:
Molecular Weight:
115.17
Beilstein:
79851
EC Number:
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

96%

form

liquid

optical activity

[α]19/D −49.5°, c = 5 in methanol

reaction suitability

reaction type: solution phase peptide synthesis

refractive index

n20/D 1.469 (lit.)

bp

67-69 °C/12 mmHg (lit.)

density

0.968 g/mL at 25 °C (lit.)

application(s)

peptide synthesis

storage temp.

2-8°C

SMILES string

CN1CCC[C@H]1CO

InChI

1S/C6H13NO/c1-7-4-2-3-6(7)5-8/h6,8H,2-5H2,1H3/t6-/m0/s1

InChI key

VCOJPHPOVDIRJK-LURJTMIESA-N

Looking for similar products? Visit Product Comparison Guide

Application

Precursor to phosphine ligands for catalytic asymmetric Grignard cross-coupling reactions.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

145.4 °F - closed cup

Flash Point(C)

63 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The Journal of Organic Chemistry, 48, 2195-2195 (1983)
Dan Loewenthal et al.
Archives of toxicology, 94(11), 3751-3757 (2020-07-29)
We present a simple method for chiral separation and analysis of organophosphorus nerve agents and apply it to monitor the enantioselective blood elimination kinetics of sarin in-vitro. The method is implemented in standard reverse phase LC-MS operating conditions, relieving the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service