Skip to Content
Merck
All Photos(1)

Key Documents

186511

Sigma-Aldrich

(S)-(+)-2-Pyrrolidinemethanol

97%

Synonym(s):

(S)-(+)-2-(Hydroxymethyl)pyrrolidine, L-Prolinol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H11NO
CAS Number:
Molecular Weight:
101.15
Beilstein:
79843
EC Number:
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

optical activity

[α]20/D +31°, c = 1 in toluene

refractive index

n20/D 1.4853 (lit.)

bp

74-76 °C/2 mmHg (lit.)

density

1.025 g/mL at 25 °C (lit.)

functional group

hydroxyl

SMILES string

OC[C@@H]1CCCN1

InChI

1S/C5H11NO/c7-4-5-2-1-3-6-5/h5-7H,1-4H2/t5-/m0/s1

InChI key

HVVNJUAVDAZWCB-YFKPBYRVSA-N

General description

(S)-(+)-2-Pyrrolidinemethanol, also known as (S)-(+)-prolinol is a chiral building block that is used for the synthesis of chiral organic compounds. It is also used as a chiral auxiliary in asymmetric synthesis and a chiral ligand in asymmetric catalysis.

Application

(S)-(+)-2-Pyrrolidinemethanol can be used as a starting material for the synthesis of (S)-α, α-diaryl-2-pyrrolidinemethanols. It plays an important role in this synthesis by providing chirality to the final product.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

186.8 °F - closed cup

Flash Point(C)

86 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Janine Cossy
Chemical record (New York, N.Y.), 5(2), 70-80 (2005-04-13)
The synthesis of optically active substituted piperidines has been achieved by using four different methodologies. The first one is an intramolecular nucleophilic displacement of activated alcohol moieties that was used to build up the piperidine ring of (-)-prosophylline and (-)-slaframine
Magnus Rueping et al.
Organic & biomolecular chemistry, 10(30), 6201-6210 (2012-05-16)
A highly efficient route for the synthesis of valuable 3,4-substituted chromenone derivatives by the reaction of 1,3-diketones with aldehydes in the presence of l-proline was developed. The reactions take advantage of readily available starting materials and follow a Knoevenagel condensation/Michael
Yuko Kawanami et al.
The Journal of organic chemistry, 74(20), 7908-7921 (2009-09-19)
The photochirogenesis of 2-anthracenecarboxylic acid (AC) complexed to a hydrogen-bonding template (TKS159) was investigated to obtain mechanistic information on how chirogenesis is achieved for the dimerization of AC. Complexation of AC to TKS159 leads to the shielding of one of
Patrick Bolze et al.
Organic letters, 10(17), 3753-3756 (2008-07-29)
A simple organocatalytic approach to highly attractive chiral building blocks is presented. By the reaction of beta-ketoesters with alpha,beta-unsaturated aldehydes using a chiral TMS-protected prolinol as the catalyst, optically active 5-(trialkylsilyl)cyclohex-2-enones are formed in good yields and with 98-99% ee.
Katsuhiko Moriyama et al.
The Journal of organic chemistry, 77(21), 9846-9851 (2012-10-12)
The intramolecular aminohydroxylation of N-alkenylsulfonamides proceeded under heavy metal-free conditions to give substituted prolinol derivatives in high yields. Oxone activated by catalytic Brønsted acid worked well as an electrophilic oxidant for this reaction.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service