Skip to Content
Merck
All Photos(1)

Key Documents

324450

Sigma-Aldrich

(S)-(+)-1-(2-Pyrrolidinylmethyl)pyrrolidine

96%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H18N2
CAS Number:
Molecular Weight:
154.25
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

Assay

96%

optical activity

[α]19/D +7.0°, c = 2.4 in ethanol

refractive index

n20/D 1.4871 (lit.)

bp

99-101 °C/2 mmHg (lit.)

density

0.946 g/mL at 25 °C (lit.)

SMILES string

C1CCN(C1)C[C@@H]2CCCN2

InChI

1S/C9H18N2/c1-2-7-11(6-1)8-9-4-3-5-10-9/h9-10H,1-8H2/t9-/m0/s1

InChI key

YLBWRMSQRFEIEB-VIFPVBQESA-N

Application

(S)-(+)-1-(2-Pyrrolidinylmethyl)pyrrolidine can be used as an organocatalyst in the asymmetric synthesis of different optically active organic building blocks via Aldol condensation and Mannich reactions . It is also used as a catalyst to prepare enantioselective Michael adducts by reacting alkylidene malonates with unactivated ketones.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

190.4 °F - closed cup

Flash Point(C)

88 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

(S)-(+)-1-(2-pyrrolidinylmethyl)pyrrolidine ? an effective catalyst of asymmetric synthesis
VM Tkachuk, et al.
Zhurnal Organichnoi ta Farmatsevtichnoi Khimii, 3-16 (2014)
M Guipponi et al.
Human genetics, 109(6), 569-575 (2002-01-26)
The human TPTE gene encodes a testis-specific protein that contains four potential transmembrane domains and a protein tyrosine phosphatase motif, and shows homology to the tumor suppressor PTEN/MMAC1. Chromosomal mapping revealed multiple copies of the TPTE gene present on the
Catalytic enantioselective direct Michael additions of ketones to alkylidene malonates
Betancort JM, et al.
Tetrahedron Letters, 42(27), 4441-4444 (2001)
Tatiana Leiva-Rodríguez et al.
Cell death & disease, 9(6), 626-626 (2018-05-26)
Injured neurons should engage endogenous mechanisms of self-protection to limit neurodegeneration. Enhancing efficacy of these mechanisms or correcting dysfunctional pathways may be a successful strategy for inducing neuroprotection. Spinal motoneurons retrogradely degenerate after proximal axotomy due to mechanical detachment (avulsion)
Shoujiro Ogawa et al.
Biomedical chromatography : BMC, 30(1), 29-34 (2015-01-27)
A simple liquid chromatography/electrospray ionization-tandem mass spectrometry (LC/ESI-MS/MS) method for determination of the eicosapentaenoic acid (EPA) concentration to arachidonic acid (AA) concentration ratio in human saliva has been developed. The EPA/AA ratio in serum or plasma is widely recognized as

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service