Skip to Content
Merck
All Photos(1)

Key Documents

298182

Sigma-Aldrich

Lanthanum(III) chloride

AnhydroBeads, −10 mesh, 99.9% trace metals basis

Synonym(s):

Lanthanum trichloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
LaCl3
CAS Number:
Molecular Weight:
245.26
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
grade:
for synthesis
synthesis grade
Pricing and availability is not currently available.

grade

for synthesis
synthesis grade

product line

AnhydroBeads

Assay

99.9% trace metals basis

reaction suitability

reagent type: catalyst
core: lanthanum

impurities

≤1500.0 ppm Trace Rare Earth Analysis

particle size

−10 mesh

bp

1812 °C (lit.)

mp

860 °C (lit.)

density

3.84 g/mL at 25 °C (lit.)

SMILES string

Cl[La](Cl)Cl

InChI

1S/3ClH.La/h3*1H;/q;;;+3/p-3

InChI key

ICAKDTKJOYSXGC-UHFFFAOYSA-K

Looking for similar products? Visit Product Comparison Guide

Legal Information

AnhydroBeads is a trademark of Sigma-Aldrich Co. LLC

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Eye Dam. 1 - Met. Corr. 1 - Skin Sens. 1

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yue Zheng et al.
mBio, 9(2) (2018-03-29)
A recent surprising discovery of the activity of rare earth metals (lanthanides) as enzyme cofactors as well as transcriptional regulators has overturned the traditional assumption of biological inertia of these metals. However, so far, examples of such activities have been
Qinyuan Xu et al.
Bioresource technology, 289, 121600-121600 (2019-06-21)
Phosphorus reduction from wastewater is vital to mitigate eutrophication of receiving waters. In this study, discarded lignocellulose biochar loaded with lanthanum (defined as La-biochar) was applied for phosphate adsorption. Based on the design of response surface methodology, La-biochar displayed a
Y Zhao et al.
British journal of cancer, 111(1), 125-131 (2014-05-29)
Signalling events mediated by connexins and cyclooxygenase-2 (COX-2) have important roles in bystander effects induced by ionising radiation. However, whether these proteins mediate bystander effects independently or cooperatively has not been investigated. Bystander normal human fibroblasts were cocultured with irradiated
Tawnya G Flick et al.
Journal of the American Society for Mass Spectrometry, 23(11), 1885-1895 (2012-09-06)
Addition of 1.0 mM LaCl(3) to aqueous ammonium acetate solutions containing proteins in their folded native forms can result in a significant increase in the molecular ion charging obtained with electrospray ionization as a result of cation adduction. In combination
T Wu et al.
American journal of physiology. Heart and circulatory physiology, 301(1), H69-H78 (2011-04-05)
The cochlear lateral wall generates the endocochlear potential (EP), which creates a driving force for the hair cell transduction current and is essential for normal hearing. Blood flow at the cochlear lateral wall is critically important for maintaining the EP.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service