Skip to Content
Merck
All Photos(3)

Documents

211605

Sigma-Aldrich

Lanthanum(III) chloride hydrate

99.9% trace metals basis

Synonym(s):

Lanthanum trichloride hydrate, Trichlorolanthanum hydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
LaCl3 · xH2O
CAS Number:
Molecular Weight:
245.26 (anhydrous basis)
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

Assay

99.9% trace metals basis

form

powder, crystals or chunks

composition

Degree of hydration, ~6

reaction suitability

reagent type: catalyst
core: lanthanum

impurities

≤1500.0 ppm Trace Rare Earth Analysis

mp

92-93 °C (lit.)

SMILES string

O.Cl[La](Cl)Cl

InChI

1S/3ClH.La.H2O/h3*1H;;1H2/q;;;+3;/p-3

InChI key

NUXZAAJDCYMILL-UHFFFAOYSA-K

Looking for similar products? Visit Product Comparison Guide

General description

Lanthanum(III) chloride hydrate is a highly water-soluble crystalline solid widely used in catalytic processes and as a dopant in the production of phosphors and ceramics.

Application

Lanthanum(III) chloride hydrate can be used:
  • As a precursor to synthesize La-based nanostructures.
  • As a dopant to fabricate ZnO nanoparticles with enhanced optical particles.
  • To synthesize Fe–La composite (hydr)oxides for arsenic removal from water.
  • Tofabricate lanthanum hexaboride (LaB6) powders forultra-high-temperature ceramics.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mohammad Hasan Moshafi et al.
International journal of nanomedicine, 14, 10137-10146 (2020-01-11)
In this study, for the first time, new nanoparticles of La3+/α-Al2O3 were synthesized with the ultrasonic-assisted hydrothermal method in the presence of honey as an eco-friendly and natural reagent. The as-synthesized La3+/α-Al2O3 nanoparticles were characterized using scanning electron microscopy (SEM)
Feng Luo et al.
Environmental science and pollution research international, 28(4), 4342-4351 (2020-09-19)
Lanthanum molybdate/magnetite (M-La2(MoO4)3) with various LaCl3/Fe3O4 mass ratios was synthesized and optimized for selective phosphate removal from wastewater. M-La2(MoO4)3 (2:1) was selected on the basis of phosphate sorption capacity for further experiments and characterized by a variety of methods. The
Haotian Hao et al.
Water research, 155, 1-11 (2019-03-04)
Effectively eutrophication control and phosphate recovery have received increasing attention in recent years. In this study, a regenerable magnetic NaLa(CO3)2/Fe3O4 composites (MLC) which includes a novel phosphate-binding lanthanum species NaLa(CO3)2 hybridized with Fe3O4 nanoparticle was developed through a modified solvothermal
Enhanced arsenate removal by novel Fe--La composite (hydr) oxides synthesized via coprecipitation
Wei Zhang, et al.
Chemical Engineering Journal, 251, 69-79 (2014)
You Wu et al.
Journal of environmental management, 231, 370-379 (2018-10-28)
The requirement to the phosphorus (P) emission from wastewater treatment plants (WWTPs) is becoming increasingly strict, which makes an advanced treatment for the low-concentration phosphate removal from secondary effluents indispensable. In present work, hydrated lanthanum (La) oxide-modified diatomite composites (La-diatomite)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service