Skip to Content
Merck
All Photos(3)

Key Documents

294020

Sigma-Aldrich

Scandium(III) oxide

powder, 99.995% trace rare earth metals basis

Synonym(s):

Scandia, Scandium trioxide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Sc2O3
CAS Number:
Molecular Weight:
137.91
EC Number:
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

99.995% trace rare earth metals basis

form

powder

reaction suitability

core: scandium
reagent type: catalyst

SMILES string

O=[Sc]O[Sc]=O

InChI

1S/3O.2Sc

InChI key

HYXGAEYDKFCVMU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

A high melting white solid used in high-temperature systems (for its resistance to heat and thermal shock), electronic ceramics, and glass composition.
Suitable for vacuum deposition applications

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R Soulard et al.
Optics express, 18(11), 11173-11180 (2010-07-01)
We report on the measurements of near-UV excited-state absorption (ESA) spectra and refractive index changes (RICs) in the two ytterbium doped laser crystals Yb:Lu2O3 and Yb:Sc2O3. ESA is assigned to ligand-to-metal charge transfer (LMCT) absorption transitions and RICs to the
[Industrial hygiene in scandium oxide manufacture].
B G Feĭgin
Gigiena truda i professional'nye zabolevaniia, (4)(4), 39-40 (1985-04-01)
Ya-Wen Zhang et al.
The journal of physical chemistry. B, 109(39), 18324-18331 (2006-07-21)
Nanostructured scandium hydrous oxides were hydrothermally synthesized at 180 degrees C for 18 h, using NaOH, NH(4)OH, and KOH as the bases. They were characterized by means of X-ray diffraction (XRD), transmission electron microscopy (TEM), N2 adsorption, thermogravimetry and differential
Yan-Xia Zhao et al.
Physical chemistry chemical physics : PCCP, 13(21), 10084-10090 (2011-04-28)
The Sc(3)O(6)(-) cluster anions were produced by laser ablation and studied by reaction with n-butane in a fast flow reactor and by photoelectron spectroscopy. The reactivity experiments indicated that one Sc(3)O(6)(-) cluster can activate two n-butane molecules consecutively with rate
H M T U Herath et al.
Journal of materials science. Materials in medicine, 16(11), 1061-1065 (2006-01-03)
Biocompatibility is a pre-requisite for all biomaterials used for medical application. During the last two decades significant advances have been made in the development of novel materials and selection and use of these materials has been directly dependent upon their

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service