Skip to Content
Merck
All Photos(2)

Documents

248800

Sigma-Aldrich

Samarium(III) chloride hexahydrate

≥99%

Synonym(s):

Samarium trichloride hydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
SmCl3 · 6H2O
CAS Number:
Molecular Weight:
364.81
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

Assay

≥99%

form

powder and chunks

reaction suitability

reagent type: catalyst
core: samarium

density

2.383 g/mL at 25 °C (lit.)

SMILES string

[H]O[H].[H]O[H].[H]O[H].[H]O[H].[H]O[H].[H]O[H].Cl[Sm](Cl)Cl

InChI

1S/3ClH.6H2O.Sm/h3*1H;6*1H2;/q;;;;;;;;;+3/p-3

InChI key

TXVNDKHBDRURNU-UHFFFAOYSA-K

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Markus Martincic et al.
Physical chemistry chemical physics : PCCP, 17(47), 31662-31669 (2015-11-12)
The endohedral functionalization of carbon nanotubes with both organic and inorganic materials allows the development of tailored functional hybrids whose properties benefit from the synergistic effects of the constituent compounds. Bulk filling of carbon nanotubes (CNTs) results in samples that

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service